Skip to Content
Merck
All Photos(6)

Key Documents

HPA013143

Sigma-Aldrich

Anti-CST3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cystatin-3, Anti-Cystatin-C precursor, Anti-Gamma-trace, Anti-Neuroendocrine basic polypeptide, Anti-Post-gamma-globulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SSPGKPPRLVGGPMDASVEEEGVRRALDFAVGEYNKASNDMYHSRALQVVRARKQIVAGVNYFLDVELGRTTCTKTQPNLDNCPFHDQPHLKRKAFCSFQIYAVPWQGTMTLSKSTCQDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CST3(1471)

General description

Cystatin-C precursor (CST3) belongs to the cystatin family (type2) of proteins and contains 120 amino acids. The gene CST3 is located on human chromosome 20p11.2. CST3 is expressed in many tissues and is present in all body fluids.

Immunogen

Cystatin-C precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cystatin-C precursor (CST3) is an inhibitor of lysosomal cysteine proteases and several cathepsins like cathepsin S, a lysosomal enzyme. It has been shown that CST3 is a marker of glomerular filtration and diabetic kidney disease. In presence of pathological conditions, it forms amyloid fibrils in brain arteries. Mutations in the gene encoding CST3 have been shown to be associated with a form of amyloidosis and age-related macular degeneration.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71715

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikola Sekulovski et al.
Biology of reproduction, 100(2), 398-408 (2018-10-18)
Endometriosis causes severe chronic pelvic pain and infertility. We have recently reported that niclosamide treatment reduces growth and progression of endometriosis-like lesions and inflammatory signaling (NF${\rm \small K}$B and STAT3) in a mouse model. In the present study, we examined
Michael E March et al.
Nature communications, 12(1), 1827-1827 (2021-03-25)
Hereditary cystatin C amyloid angiopathy is a dominantly inherited disease caused by a leucine to glutamine variant of human cystatin C (hCC). L68Q-hCC forms amyloid deposits in brain arteries associated with micro-infarcts, leading ultimately to paralysis, dementia and death in
Elisa Teyssou et al.
Neurobiology of aging, 99, 102-102 (2020-11-22)
ANXA11 mutations have previously been discovered in amyotrophic lateral sclerosis (ALS) motor neuron disease. To confirm the contribution of ANXA11 mutations to ALS, a large exome data set obtained from 330 French patients, including 150 familial ALS index cases and
Paraskevi L Tsiolaki et al.
FEBS letters, 589(1), 159-164 (2014-12-06)
Human cystatin C (HCC) is a low molecular weight member of the cystatin family (type2). HCC consists of 120 amino acids. Normally it is an inhibitor of cysteine proteases, but in pathological conditions it forms amyloid fibrils in brain arteries
Zahra Asefy et al.
Pakistan journal of biological sciences : PJBS, 17(4), 582-585 (2015-04-29)
Serum concentration of cystatin C a marker of glomerular filtration has been associated with Cardiovascular Disease (CVD). The aim of this study was to evaluate cystatin C as a marker of diabetic kidney disease in normoalbuminuric diabetic patients without Chronic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service