Skip to Content
Merck
All Photos(2)

Key Documents

HPA011185

Sigma-Aldrich

Anti-NEXN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-F-actin binding protein, Anti-Nelin, Anti-Nexilin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

EEERARRRAIDLEIKEREAENFHEEDDVDVRPARKSEAPFTHKVNMKARFEQMAKAREEEEQRRIEEQKLLRMQFEQREIDAALQKKREEEEEEEGSIMNGSTAEDEEQTRSGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEXN(91624)

General description

NEXN (nexilin) is an F-actin binding protein, which is muscle-specific. It resides in adherens junctions. It is predominantly expressed in heart and skeletal muscles. This gene maps to human chromosome 1p31-1p32.

Immunogen

Nexilin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NEXN (nexilin) is responsible for the stabilization of Z-disc and force generation in heart. In smooth muscle cells, its expression is suppressed in response to inflammatory cytokines. It protects vascular smooth muscle cells in injured vessels, by promoting contractility. Variations in this gene are associated with coronary artery diseases in humans. It is a part of focal adhesions, and is thought to play a role in actin-related motility. It is a part of the actin-rich structure of Listeria monocytogenes and enteropathogenic Escherichia coli, and is essential for their invasiveness and infection. Mutations in this gene are also responsible for hypertrophic cardiomyopathy (HCM), which is an inherited disorder characterized by cardiomyocyte hypertrophy, increase in fibrosis of myocardium and increased wall thickness of ventricles. NEXN might also be involved in the formation of stress fibers and focal adhesions, as well as form a part of focal adhesion signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72487

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chong Wu et al.
PloS one, 8(12), e82135-e82135 (2013-12-19)
Coronary artery disease (CAD) is the leading cause of death and disability in the world. Genome-wide association studies have implicated the importance of the genetic contribution of vascular smooth muscle cells (VSMCs) function in CAD susceptibility. The aberrant phenotypic modulation
Wei Wang et al.
Biochemical and biophysical research communications, 330(4), 1127-1131 (2005-04-13)
A new gene (GenBank Accession No. AF114264) was cloned from umbilical vein wall tissue by using RT-PCR. The gene shares high similarity to the gene encoding F-actin binding protein nexilin, so named as NELIN. A clone of 2737bp contains open
Baoyi Zhu et al.
Scientific reports, 8(1), 13025-13025 (2018-08-31)
Nexilin, encoded by the NEXN gene, is expressed in striated muscle and localizes to Z-discs, influencing mechanical stability. We examined Nexilin/NEXN in smooth muscle cells (SMCs), and addressed if Nexilin localizes to dense bodies and dense bands and whether it
Yong Zhao et al.
Sheng wu hua xue yu sheng wu wu li xue bao Acta biochimica et biophysica Sinica, 33(1), 19-24 (2002-06-08)
For searching cardiovascular-associated genes and investigating their expression profiles, human adult heart and aorta cDNA libraries were constructed, and a novel gene from adult heart cDNA library was isolated based on large-scale ESTs(expressed sequence tags) sequencing(GenBank accession number AF114264). The
H T Law et al.
Cellular microbiology, 14(7), 1097-1108 (2012-03-03)
The bacterial pathogens Listeria monocytogenes and enteropathogenic Escherichia coli (EPEC) generate motile actin-rich structures (comet tails and pedestals) as part of their infectious processes. Nexilin, an actin-associated protein and a component of focal adhesions, has been suggested to be involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service