Skip to Content
Merck
All Photos(8)

Key Documents

HPA007264

Sigma-Aldrich

Anti-SSTR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SRIF-1 antibody produced in rabbit, Anti-SS2R antibody produced in rabbit, Anti-Somatostatin receptor type 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DNFKKSFQNVLCLVKVSGTDDGERSDSKQDKSRLNETTETQRTLLNGDLQTSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SSTR2(6752)

General description

SSTR2 (Somatostatin receptor 2) is a novel multi-domain protein belonging to a family of cytoskeletal anchoring proteins. It consists of six N-terminal ankyrin repeats followed by SH3 and PDZ domains, several proline-rich regions, and a C-terminal sterile α region.

Immunogen

Somatostatin receptor type 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-SSTR2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

SSTR2 (Somatostatin receptor 2) is a regulatory peptide widely distributed in tissues. It is involved in several cellular functions including, neuroendocrine and exocrine secretions, and proliferation. In neuronal tissues, it interacts with the inhibitory G-proteins in both the chambers, presynaptic and postsynaptic, to control the synaptic responses. It has the ability to control the proliferation of both normal and neoplastic cells. It is abundantly present at the plasma membrane of several tumorous cells such as pancreatic, gastrointestinal, breast carcinoma, neuroblastomas etc. Thus, SSTR2 has been proved as a potential prognostic marker for certain malignancies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70183

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Remke et al.
Child's nervous system : ChNS : official journal of the International Society for Pediatric Neurosurgery, 29(8), 1253-1262 (2013-05-17)
Neuroectodermal tumors in general demonstrate high and dense expression of the somatostatin receptor subtype 2 (sst₂). It controls proliferation of both normal and neoplastic cells. sst₂ has thus been suggested as a therapeutic target and prognostic marker for certain malignancies.
Catherine G Tran et al.
Cancers, 14(8) (2022-04-24)
Gastroenteropancreatic neuroendocrine neoplasms (GEP NENs) are rare cancers consisting of neuroendocrine carcinomas (NECs) and neuroendocrine tumors (NETs), which have been increasing in incidence in recent years. Few cell lines and pre-clinical models exist for studying GEP NECs and NETs, limiting
Joanna Fombonne et al.
Reproductive biology and endocrinology : RB&E, 1, 19-19 (2003-03-21)
We recently reported that immature porcine Leydig cells express both somatostatin (SRIF) and SRIF receptor type-2 (sst-2) transcripts. The present study was therefore undertaken to assess whether SRIF might exert autocrine actions on these cells through sst2A receptor, one of
Martin Ullrich et al.
Endocrine-related cancer, 25(12), 993-1004 (2018-10-06)
Somatostatin receptor-targeting endoradiotherapy offers potential for treating metastatic pheochromocytomas and paragangliomas, an approach likely to benefit from combination radiosensitization therapy. To provide reliable preclinical in vivo models of metastatic disease, this study characterized the metastatic spread of luciferase-expressing mouse pheochromocytoma
Natasha Alexander et al.
Journal of pediatric hematology/oncology, 41(3), 222-227 (2018-10-20)
Alternative radiolabeled, targeted agents are being investigated for children with relapsed neuroblastoma (NB) who do not respond to I-metaiodobenzylguanidine (MIBG) therapy. (DOTA-Tyr)-octreotate targets somatostatin receptors (SSTRs), particularly SSTR2, which are expressed on NB cells. We investigated SSTR2 expression in NB

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service