Skip to Content
Merck
All Photos(1)

Key Documents

HPA006206

Sigma-Aldrich

Anti-EGR3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PILOT, Anti-early growth response 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

EPVSFHDPQGNPGLAYSPQDYQSAKPALDSNLFPMIPDYNLYHHPNDMGSIPEHKPFQGMDPIRVNPPPITPLETIKAFKDKQIHPGFGSLPQPPLTLKPIRPRKYPNRPSKTPLHERPHACPAEGCDRRFSRSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EGR3(1960)

Immunogen

early growth response 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-EGR3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Early growth response protein 3 is a protein encoded by the EGR3 gene in humans. It belongs to Egr family; possess a single intron and maps to chromosome 8 at bands p21-23. This gene is an immediate early gene that is expressed throughout the brain and is a susceptibility gene for schizophrenia (SZ). It plays a key role in the vascular endothelial growth factor (VEGF) signalling in activated endothelial cells. Polymorphism of this gene is associated with coronary artery disease (CAD) in males of the Chinese Uygur population. Its expression may play an important role in the differentiation, proliferation, metastasis and progression of gastric cancer. It may also act as a potential diagnostic marker for gastric cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70994

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukika Nishimura et al.
NeuroImage, 85 Pt 1, 527-534 (2013-08-22)
The early growth response 3 (EGR3) gene is an immediate early gene that is expressed throughout the brain and has been suggested as a potential susceptibility gene for schizophrenia (SZ). EGR3 impairment is associated with various neurodevelopmental dysfunctions, and some
Xia Li et al.
Lipids in health and disease, 13, 84-84 (2014-06-03)
Endothelial cell activation and dysfunction are the foundation of atherosclerosis, including coronary artery disease (CAD). Endothelial cell activation is mediated by the level of gene transcription. Early growth response 3 (Egr3) is a critical determinant of vascular endothelial growth factor
Fei Liao et al.
Journal of molecular histology, 44(4), 463-468 (2013-03-06)
The early growth response (EGR) family has a highly conserved DNA-binding domain and encodes zinc finger proteins, which show suppressive effects on tumour growth. However, the expression and significance of EGR3 in gastric cancer are still unknown. In this study
Qi-Lan Ning et al.
Yi chuan = Hereditas, 34(3), 307-314 (2012-03-20)
Previous studies showed that EGR3 gene located in chromosome 8p21.3 was involved in the etiology of schizophrenia. However, the finding failed to be replicated in several case-control studies. To investigate the genetic role of the EGR3 gene in Chinese psychiatric
S Patwardhan et al.
Oncogene, 6(6), 917-928 (1991-06-01)
We have previously described two cellular immediate-early genes, Egr-1 (mouse) and EGR2 (human) that encode zinc finger proteins. Here we report the characterization of a new member of the Egr family referred to as EGR3 (human). This cDNA clone was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service