Skip to Content
Merck
All Photos(1)

Key Documents

S1061

Sigma-Aldrich

Stat-Matic plate washer

Model II

Synonym(s):

Plate washer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.22

sterility

non-sterile

manufacturer/tradename

Tricontinent 0980

size

96 wells

volumes dispensed

100, 200, 250 or 300 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Unit includes 500 mL plastic bottle with 288 mm cap assembly with bottle inlet tubing and Luer fitting, 3 dispense volume spacers, one 30 in. long Tygon remote inlet tubing.
Fast, convenient, accurate filling or washing of microplates from an external reservoir. All inert polypropylene-and-glass construction with a PTFE plunger. Set manifold by inserting spacers for 100, 200, or 250 μL and dispense into eight wells at a time (spacing set for 96 well plates).

Features and Benefits

  • Eight-port manifold, designed for 96-well microplates, delivers fluids in volumes of 100, 200, 250, or 300 μL with well-to-well accuracy of 5% or better.
  • Constructed of polypropylene, PTFE, glass, and ceramics, there is no exposed glass that can break and requires minimal maintenance.
  • Compatible with most reagents and chemicals and designed so that even protein-based reagents won′t cause sticking.
  • Filtered air vent keeps liquids free of contaminants.
  • Priming line allows recirculation into the dispenser port without removing the bottle.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibiao Xie et al.
Cell death & disease, 12(1), 1-1 (2021-01-09)
TP53 is a classic tumor suppressor, but its role in kidney cancer remains unclear. In our study, we tried to explain the role of p53 in kidney cancer through the p53-related enhancer RNA pathway. Functional experiments were used to explore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service