Skip to Content
Merck
All Photos(5)

Documents

HPA005670

Sigma-Aldrich

Anti-DLX5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Homeobox protein DLX-5 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RRVPSIRSGDFQAPFQTSAAMHHPSQESPTLPESSATDSDYYSPTGGAPHGYCSPTSASYGKALNPYQYQYHGVNGSAGSYPAKAYADYSYASSYHQYGGAYNRVPSATNQPEKEVTEPEVRMVNGKPKKVRKP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLX5(1749)

General description

Distal-less homeobox 5 (DLX5) belongs to the Dlx gene family which encodes a group of developmental regulatory proteins and transcription factors that are expressed primarily in embryonic development.

Immunogen

Homeobox protein DLX-5 recombinant protein epitope signature tag (PrEST)

Application

Anti-DLX5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Distal-less homeobox 5 (DLX5) plays a role as a mediator in immediate-early bone morphogenic protein (BMP)-induced runt-related transcription factor 2 (RUNX2) (up regulated by DLX2) expression and as a responsive transcriptional activator essential for osteoblast differentiation. Msh homeobox 2 (MSX2) and DLX5 bind to the promoters of osteoblast markers, thus helping to carry out transcription. It is involved as a positive regulator of both chondrogenesis and chondrocyte hypertrophy in the endochondral skeleton. DLX5 stimulates Alkaline phosphatase (ALPL) promoter activity in a RUNX2-independent manner during osteoblast differentiation and also stimulates SP7 transcription factor (SP7) promoter activity. This is done by binding to the homeodomain-response element of the ALPL and SP7 promoter. It promotes cell proliferation by binding and up-regulating the MYC promoter activity. For this, DLX5 requires the 5′-TAATTA-3′ consensus sequence to bind to the DNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86924

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Marques et al.
PLoS genetics, 12(9), e1006307-e1006307 (2016-09-14)
The acrofacial dysostoses (AFD) are a genetically heterogeneous group of inherited disorders with craniofacial and limb abnormalities. Rodriguez syndrome is a severe, usually perinatal lethal AFD, characterized by severe retrognathia, oligodactyly and lower limb abnormalities. Rodriguez syndrome has been proposed
Rachel D Mullen et al.
Frontiers in endocrinology, 13, 916173-916173 (2022-08-02)
Dlx5 and Dlx6 encode distal-less homeodomain transcription factors that are present in the genome as a linked pair at a single locus. Dlx5 and Dlx6 have redundant roles in craniofacial, skeletal, and uterine development. Previously, we performed a transcriptome comparison
Jinfei Xu et al.
The Journal of biological chemistry, 284(31), 20593-20601 (2009-06-06)
The human DLX homeobox genes, which are related to Dll (Drosophila distal-less gene), encode transcription factors that are expressed primarily in embryonic development. Recently, DLX5 was reported to act as an oncogene in lymphomas and lung cancers, although the mechanism
Kamal Bouhali et al.
Human molecular genetics, 20(13), 2642-2650 (2011-04-21)
Primary ovarian insufficiency (POI) is characterized by the loss of ovarian function before the age of 40 in humans. Although most cases of POI are idiopathic, many are familial, underlying a genetic origin of the disease. Mutations in genes involved
Mi-Hye Lee et al.
The Journal of biological chemistry, 280(42), 35579-35587 (2005-08-24)
Two major isoforms of the Runx2 gene are expressed by alternative promoter usage: Runx2 type I (Runx2-I) is derived from the proximal promoter (P2), and Runx2 type II (Runx2-II) is produced by the distal promoter (P1). Our previous results indicate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service