Skip to Content
Merck
All Photos(7)

Key Documents

HPA003418

Sigma-Aldrich

Anti-GZMB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C11 antibody produced in rabbit, Anti-CTLA-1 antibody produced in rabbit, Anti-CTSGL1 antibody produced in rabbit, Anti-Cathepsin G-like 1 antibody produced in rabbit, Anti-Cytotoxic T-lymphocyte proteinase 2 antibody produced in rabbit, Anti-Fragmentin-2 antibody produced in rabbit, Anti-Granzyme B precursor antibody produced in rabbit, Anti-Granzyme-2 antibody produced in rabbit, Anti-HLP antibody produced in rabbit, Anti-Human lymphocyte protein antibody produced in rabbit, Anti-Lymphocyte protease antibody produced in rabbit, Anti-SECT antibody produced in rabbit, Anti-T-cell serine protease 1-3E antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TRAVQPLRLPSNKAQVKPGQTCSVAGWGQTAPLGKHSHTLQEVKMTVQEDRKCESDLRHYYDSTIELCVGDPEIKKTSFKGDSGGPLVCNKVAQGIVSYGRNNGMPPRACTKVSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GZMB(3002)

General description

Granzyme B is a serine protease encoded by the GZMB gene in humans. It is secreted from cytotoxic lymphocytes within the rheumatoid joint.

Immunogen

Granzyme B precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Granzyme B inhibitor is found in human small intestine. During prolonged inflammation, the serine protease granzyme B circulating level is increased and is implicated in the pathogenesis of several chronic inflammatory diseases. It is may have a possible role in type 2 diabetes mellitus (T2DM).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70654

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hala O El Mesallamy et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 34(3), 179-186 (2013-11-08)
Chronic inflammation and insulin resistance form hallmarks of type 2 diabetes mellitus (T2DM). An increased circulating level of the serine protease granzyme B (GzmB) is observed during prolonged inflammation and is implicated in the pathogenesis of several chronic inflammatory diseases.
Maria Pino et al.
PLoS pathogens, 15(10), e1008081-e1008081 (2019-10-19)
Lymph nodes (LN) and their resident T follicular helper CD4+ T cells (Tfh) are a critical site for HIV replication and persistence. Therefore, optimizing antiviral activity in lymphoid tissues will be needed to reduce or eliminate the HIV reservoir. In
C J Froelich et al.
Journal of immunology (Baltimore, Md. : 1950), 151(12), 7161-7171 (1993-12-15)
Cartilage degradation, a hallmark of rheumatoid arthritis, is attributed to serine and metalloproteases secreted by neutrophils, synovial lining cells, macrophages, and chondrocytes. A large proportion of synovial fluid lymphocytes contains the granule-associated serine proteases granzymes A and B. We report
V-M Pohjanen et al.
International journal of immunopathology and pharmacology, 26(4), 897-905 (2013-12-21)
The objective of this study was to assess the expression of protease inhibitor 9, a granzyme B inhibitor, in human small intestine, and to evaluate its cytoprotective role in the celiac disease of children. Twelve subjects with untreated celiac disease
Steffen Wagner et al.
International journal of cancer, 138(9), 2263-2273 (2015-12-15)
Human papillomavirus (HPV)-related squamous cell carcinoma of the oropharynx (OSCC) are clinical and biological distinct from their HPV-unrelated counterparts. Patients with HPV-related OSCC display improved prognosis and therefore we investigated possible immune cell infiltrations associated with this tumor phenotype. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service