Skip to Content
Merck
All Photos(7)

Key Documents

HPA002531

Sigma-Aldrich

Anti-IRF8 antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

ICSBP, Interferon consensus sequence-binding protein, Interferon regulatory factor 8

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRF8(3394)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Interferon regulatory factor 8 recombinant protein epitope signature tag (PrEST)

Sequence
PDFEEVTDRSQLDISEPYKVYRIVPEEEQKCKLGVATAGCVNEVTEMECGRSEIDELIKEPSVDDYMGMIKRSPSPPEACRSQLLPDWWAQQPSTGVPLVTGYTTYDAHHSAFSQMVISFYY

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IRF8 (interferon regulatory factor 8) plays a vital role in regulating the proliferation and differentiation of hematopoietic progenitor cells. Deficiency of IRF8 causes chronic myelogenous leukemia (CML)-like syndrome that proves its vital role in the development of myeloid cells. It forms complex with other members of the IRF family and IFN (interferon)-stimulated response element (ISRE) to regulate type I IFNs stimulated gene expression, namely IFN-α and IFN-β. It has also been reported that IRF8 has ability to inhibit DNA-binding activity of IRF9.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86219

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Bovolenta et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(11), 5046-5050 (1994-05-24)
Interferon (IFN) consensus sequence binding protein (ICSBP) is a transcription factor expressed mostly in the cells of the immune system. ICSBP belongs to the IFN regulatory factor (IRF) family, which also includes IRF-1, IRF-2, and the IFN-alpha-stimulated gene factor 3
T Holtschke et al.
Cell, 87(2), 307-317 (1996-10-18)
Interferon consensus sequence binding protein (ICSBP) is a transcription factor of the interferon (IFN) regulatory factor (IRF) family. Mice with a null mutation of ICSBP exhibit two prominent phenotypes related to previously described activities of the IRF family. The first
Tomohiko Tamura et al.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research, 22(1), 145-152 (2002-02-16)
Interferon (IFN) consensus sequence binding protein (ICSBP)/IFN regulatory factor (IRF)-8 is an IFNgamma-inducible transcription factor of the IRF family and regulates transcription through multiple target DNA elements, such as IFN-stimulated response element (ISRE), Ets/IRF composite element, and IFN-gamma activation site
Correction: Reprogramming anchorage dependency by adherent‑to‑suspension transition promotes metastatic dissemination.
Hyunbin D Huh et al.
Molecular cancer, 22(1), 135-135 (2023-08-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service