Skip to Content
Merck
All Photos(3)

Documents

HPA002185

Sigma-Aldrich

Anti-CUL5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CUL-5 antibody produced in rabbit, Anti-Cullin-5 antibody produced in rabbit, Anti-VACM-1 antibody produced in rabbit, Anti-Vasopressin-activated calcium-mobilizing receptor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

FSLMDKVPNGIEPMLKDLEEHIISAGLADMVAAAETITTDSEKYVEQLLTLFNRFSKLVKEAFQDDPRFLTARDKAYKAVVNDATIFKLELPLKQKGVGLKTQPESKCPELLANYCDMLLRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUL5(8065)

General description

Cullin-5 (CUL5) is a vasopressin-activated calcium-mobilizing receptor 1 and a component of cullin-RING ubiquitin ligase (CRL) complex and elongin-cullin-SOCS box (ECS) E3 ubiquitin ligase. It comprises an N-terminal and C-terminal domain. The CUL5 gene is mapped to human chromosome 11q22.3.

Immunogen

Cullin-5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CUL5 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

CUL5 (cullin 5), a tumor suppressor gene, belongs to the family of cullin genes. It is involved in the cell cycle regulation as well as in tumor progression. During cortical development, CUL5 forms complex with SOCS (suppressors of cytokine signaling) proteins to regulate neuron migrations. It may function as a tumor suppressor in hepatocellular carcinoma (HCC) tissues. It has been reported that CUL5 conjugated with heat shock protein 90 (HSP90) participates in the ubiquitination and degradation of the protein kinase client, ERBB2 (erb-b2 receptor tyrosine kinase 2), followed by HSP90 inhibition.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85185

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John D Dickinson et al.
International journal of molecular medicine, 17(5), 769-778 (2006-04-06)
This work describes the identification and impact of somatic genomic abnormalities in human chronic lymphocytic leukemia (CLL). Using molecular cytogenetics (FISH) and G-banding cytogenetic analysis, chromosome abnormalities were detected in 37 of 46 (80.4%) CLL patients. 13q14 deletion was the
Chaoqun Ma et al.
IUBMB life, 65(12), 1026-1034 (2013-12-18)
MicroRNAs (miRNAs) are small, non-coding RNAs that participate in the regulation of gene expression. In this study, we demonstrate that miR-7 was downregulated in hepatocellular carcinoma (HCC) tissues compared to adjacent non-tumor tissue. Over-expression of miR-7 in QGY-7703 and HepG2
Shuhua Zheng et al.
Aging, 12(17), 17380-17392 (2020-09-16)
Glioblastoma multiforme (GBM) is the deadliest type of brain tumor. The median survival time for patients with GBM is only 15 months, even following maximal surgical resection and chemotherapy and radiation therapy. A genetic biomarker could enable a paradigm shift
Rahul S Samant et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(18), 6834-6839 (2014-04-25)
The molecular chaperone heat shock protein 90 (HSP90) is required for the activity and stability of its client proteins. Pharmacologic inhibition of HSP90 leads to the ubiquitin-mediated degradation of clients, particularly activated or mutant oncogenic protein kinases. Client ubiquitination occurs
Feng Gao et al.
American journal of translational research, 12(2), 618-632 (2020-03-21)
Cullin-5 (CUL5), a scaffold protein in active cullin-RING ubiquitin ligase (CRL) complexes, is a member of the cullin family of proteins. The CUL5-type ubiquitin ligase can target multiple proteins involved in ubiquitination and proteasome degradation. CUL5 plays positive roles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service