Skip to Content
Merck
All Photos(7)

Key Documents

HPA002018

Sigma-Aldrich

Anti-CD36 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD36 antigen antibody produced in rabbit, Anti-FAT antibody produced in rabbit, Anti-Fatty acid translocase antibody produced in rabbit, Anti-GP-IIIB antibody produced in rabbit, Anti-GP-IV antibody produced in rabbit, Anti-Glycoprotein IIIb antibody produced in rabbit, Anti-Leukocyte differentiation antigen CD36 antibody produced in rabbit, Anti-PAS IV antibody produced in rabbit, Anti-PAS-4 protein antibody produced in rabbit, Anti-Platelet collagen receptor antibody produced in rabbit, Anti-Platelet glycoprotein 4 antibody produced in rabbit, Anti-Platelet glycoprotein IV antibody produced in rabbit, Anti-Thrombospondin receptor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

VVLEEGTIAFKNWVKTGTEVYRQFWIFDVQNPQEVMMNSSNIQVKQRGPYTYRVRFLAKENVTQDAEDNTVSFLQPNGAIFEPSLSVGTEADNFTVLNLAVAAASHIYQNQFVQMILNSLINKSKSSMFQVRTLREL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD36(948)

General description

CD36 is a major platelet membrane glycoprotein IV.

Immunogen

Platelet glycoprotein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CD36 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CD36 is regulated by Csk homologous kinase (Chk), a tyrosine kinase, in thrombin stimulated platelets. CD36 interacts with fibrinogen-liganded integrin aIIbβ3 (GP IIb/IIIa) to stabilize aggregation and activation of platelet. During platelet activation, it identifies and binds to the quaternary (polymeric) structure of collagen.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77481

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna S Enke et al.
BMC cancer, 24(1), 65-65 (2024-01-13)
Recently, we introduced Stroma-AReactive-Invasion-Front-Areas (SARIFA) as a novel hematoxylin-eosin (H&E)-based histopathologic prognostic biomarker for various gastrointestinal cancers, closely related to lipid metabolism. To date, no studies on SARIFA, which is defined as direct tumor-adipocyte-interaction, beyond the alimentary tract exist. Hence
Natascia Marino et al.
NPJ breast cancer, 6, 50-50 (2020-10-22)
Histologically normal tissue adjacent to the tumor can provide insight of the microenvironmental alterations surrounding the cancerous lesion and affecting the progression of the disease. However, little is known about the molecular changes governing cancer initiation in cancer-free breast tissue.
B Kehrel et al.
Blood, 91(2), 491-499 (1998-01-15)
Simple collagen-related peptides (CRPs) containing a repeat Gly-Pro-Hyp sequence are highly potent platelet agonists. Like collagen, they must exhibit tertiary (triple-helical) and quaternary (polymeric) structure to activate platelets. Platelet signaling events induced by the peptides are the same as most
N N Tandon et al.
The Journal of biological chemistry, 264(13), 7576-7583 (1989-05-05)
The role of glycoprotein IV (GPIV) in platelet activation processes has been examined by several different approaches: (i) Fab fragments of a monospecific polyclonal antibody to purified platelet GPIV (approximately 20 micrograms/ml) completely inhibited platelet shape change, aggregation, and secretion
Melissa M Thomas et al.
Physiological reports, 2(9), doi:10-doi:10 (2014-09-24)
Short-term consumption of a high-fat diet (HFD) can result in an oxidative shift in adult skeletal muscle. However, the impact of HFD on young, growing muscle is largely unknown. Thus, 4-week-old mice were randomly divided into sedentary HFD (60% kcal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service