Skip to Content
Merck
All Photos(1)

Key Documents

118R-1

Sigma-Aldrich

CD38 (SP149) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP149, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (118R-14)
vial of 0.5 mL concentrate (118R-15)
bottle of 1.0 mL predilute (118R-17)
vial of 1.0 mL concentrate (118R-16)
bottle of 7.0 mL predilute (118R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

bone marrow, lymph node, plasma cell myeloma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... CD38(952)

General description

CD38 molecule is a 46-kDa type-II transmembrane glycoprotein with a short N-terminalcytoplasmic tail (20 amino acids) and a long extracellular domain (256 amino acids). Itis present on many hematopoietic cells, especially plasma cells, and in some solid tumortissues. CD38 is one of the early expressed markers of mature naive B-cell activation and it isuseful in identifying functional mature B-lymphocyte subsets.

Quality


IVD

IVD

IVD

RUO

Linkage

CD38 Positive Control Slides, Product No. 118S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavius Martin et al.
Nature reviews. Immunology, 2(5), 323-335 (2002-05-30)
Recent advances in genomics and proteomics, combined with the facilitated generation and analysis of transgenic and gene-knockout animals, have revealed new complexities in classical biological systems, including the B-cell compartment. Studies on an 'old', but poorly characterized, B-cell subset--the naive
Leoncini, L et al. Burkitt lymphoma. In Swerdlow SH, et al. eds. WHO Classification of Tumours of Haematopoietic and Lymphoid Tissues. IARC, WHO Press, Lyon, France, 2008; 262-264.
Leoncini, L
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 262-264 (2008)
Kikkeri N Naresh et al.
British journal of haematology, 154(6), 770-776 (2011-07-02)
Distinguishing Burkitt lymphoma (BL) from B cell lymphoma, unclassifiable with features intermediate between diffuse large B-cell lymphoma (DLBCL) and BL (DLBCL/BL), and DLBCL is challenging. We propose an immunohistochemistry and fluorescent in situ hybridization (FISH) based scoring system that is
Fabio Malavasi et al.
Physiological reviews, 88(3), 841-886 (2008-07-16)
The membrane proteins CD38 and CD157 belong to an evolutionarily conserved family of enzymes that play crucial roles in human physiology. Expressed in distinct patterns in most tissues, CD38 (and CD157) cleaves NAD(+) and NADP(+), generating cyclic ADP ribose (cADPR)
Sandeep S Dave et al.
The New England journal of medicine, 354(23), 2431-2442 (2006-06-09)
The distinction between Burkitt's lymphoma and diffuse large-B-cell lymphoma is crucial because these two types of lymphoma require different treatments. We examined whether gene-expression profiling could reliably distinguish Burkitt's lymphoma from diffuse large-B-cell lymphoma. Tumor-biopsy specimens from 303 patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service