Skip to Content
Merck
All Photos(1)

Documents

H9661

Sigma-Aldrich

Hepatocyte Growth Factor human

HGF, recombinant, expressed in NSO cells, suitable for cell culture

Synonym(s):

HGF, Hepatopoietin A, Scatter Factor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

20-40 ng/mL

quality

endotoxin tested

mol wt

70-80 kDa by SDS-PAGE (non-reducing)

packaging

pkg of 5 and 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

<1.0 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HGF(3082)

Looking for similar products? Visit Product Comparison Guide

General description

Recombinant, human hepatocyte growth factor (HGF) is produced from the 728 amino acid residue variant of the human pre-pro-HGF protein 6 and expressed in a mouse myeloma cell line NSO.The HGF gene is mapped to human chromosome 7q21.1. The encoded protein is a mesenchymal-fibroblast derived cytokine.

Application

Hepatocyte Growth Factor human has been used as a growth factor cocktail to suspend rat fibrogen/thrombin cells. It has also been used to study the tyrosine phosphorylation of MET (MET proto-oncogene, receptor tyrosine kinase) in myxofibrosarcoma cell line.

Biochem/physiol Actions

HGF′s major function is to induce in vitro hepatocytes proliferation. HGF is also essential for cell migration and angiogenesis. Itis associated with embryonic development and tissue homeostasis. It is known to mediate placental trophoblast invasion, which is essential for normal fetal growth. Abnormal and defective HGF causes preeclampsia due to placental insufficiency. HGF exhibits its multifunctions through c-MET (proto-oncogene tyrosine kinase receptor). HGF is known to induce human umbilical vein endothelial cell proliferation and vasculogenic mimicry migration in gastric cancer cells, thus serving as a good target for cancer therapy. Upregulation of the HGF gene is observed in advanced stages of cancer, such as pancreatic adenocarcinoma.
Hepatocyte growth factor (HGF) is a potent mitogen for hepatocytes and a variety of cell types including endothelial and epithelial cells, melanocytes, and keratinocytes. It also mediates epithelial morphogenesis. HGF is the ligand for the tyrosine kinase receptor encoded by the met proto-oncogene.

Other Notes

The NSO-expressed recombinant HGF is greater than 95% heterodimeric HGF. The insect cell line-expressed recombinant HGF is a mixture of single chain HGF (predominant) with some heterodimeric HGF. Single chain HGF and heterodimeric HGF are equally active in in vitro assays.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 50 μg of bovine serum albumin per 1μg of cytokine.

Analysis Note

The biological activity is measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation between HGF/c-Met and Notch1 signaling pathways in human gastric cancer cells
Huang KH, et al.
Oncology Reports, 40(1), 294-302 (2018)
Dual inhibition of Hedgehog and c-Met pathways for pancreatic cancer treatment
Rucki AA, et al.
Molecular Cancer Therapeutics (2017)
Prognostic implication of MET overexpression in myxofibrosarcomas: an integrative array comparative genomic hybridization, real-time quantitative PCR, immunoblotting, and immunohistochemical analysis
Lee JC, et al.
Modern Pathology, 23(10), 1379-1379 (2010)
HGF derived from cancer-associated fibroblasts promotes vascularization in gastric cancer via PI3K/AKT and ERK1/2 signaling
Ding X, et al.
Oncology Reports (2018)
Analysis of hepatocyte growth factor immunostaining in the placenta of HIV-infected normotensive versus preeclamptic pregnant women
Cele SB,et al.
Eur. J. Obstet. Gyn. Rep. Biol. (2018)

Articles

Hepatocyte growth factor (HGF) is a potent mitogen for hepatocytes and a variety of other cells, including endothelial and epithelial cells, melanocytes, and keratinocytes.

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service