Skip to Content
Merck
All Photos(3)

Key Documents

E5036

Sigma-Aldrich

Epidermal Growth Factor Protein, human

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

Epidermal Growth Factor human, EGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

0.08-0.8 ng/mL EC50

mol wt

~6 kDa

packaging

pkg of 200 and 500 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

≤1 EU/μg Endotoxin

color

white

solubility

water: soluble 0.190-0.210, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H]%12N(CCC%12)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N

InChI

1S/C270H401N73O83S7/c1-24-134(19)217(262(420)293-112-201(355)298-161(69-74-205(359)360)229(387)301-160(45-35-82-286-269(279)280)228(386)333-192(119-428)255(413)307-162(68-73-198(274)352)230(388)316-176(94-141-54-64-150(350)65-55-141)241(399)304-159(44-34-

InChI key

GVUGOAYIVIDWIO-UFWWTJHBSA-N

Gene Information

human ... EGF(1950)

Looking for similar products? Visit Product Comparison Guide

General description

Epidermal Growth Factor (EGF) is a small mitogenic polypeptide (∼6 kDa), which is present in many mammalian species and is distributed throughout a wide number of tissues and body fluids. Human EGF is identical to β-urogastrone, a polypeptide which was recognized and isolated on the basis of its ability to inhibit gastric acid secretion. EGF is a member of a growth factor family, which is characterized by the presence of 6 conserved cysteine motifs that form three disulfide bonds. The location of 3 intrachain disulfide bonds in recombinant human EGF is identical to that of mouse EGF. EGF is homologous to a sequence contained in a 19 kDa protein of vaccinia virus, which appears to utilize the EGF receptor to gain entry into cell[1]† EGF is mitogenic for a variety of epidermal and epithelial cells, including fibroblasts, glial cells, mammary epithelial cells, vascular and corneal endothelial cells, bovine granulosa, rabbit chondrocytes, HeLa cells, and SV40-3T3 cells.
Epidermal growth factor (EGF) is synthesized in Henle′s loop and the distal convoluted tubule in the kidney. It is also produced in salivary glands and duodenum.[2]

Application

Epidermal Growth Factor, human, animal component free has been used:
  • as a supplement in in LHC-8 medium to culture liver cell lines [3]
  • in the fetal bovine serum (FBS)-Dulbecco′s modified essential medium (DMEM) /F12 medium for primary culture of human glioma cells[4]
  • as an additive in the conditional medium of normal fibroblasts (NFs) to study its effect on the migration and invasion of endometrial cancer (EC) cells[5]
  • as a component in tumorsphere medium[6]

Biochem/physiol Actions

Epidermal growth factor (EGF) helps to induce cell growth, proliferation and differentiation. It participates in the repairing of renal tissues in the kidney. It promotes the reabsorption of magnesium with the help of the transient receptor potential cation channel 6 (TRMP6). EGF is known to participate in the pathophysiology of drug-induced renal magnesium loss.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epidermal growth factor and its influencing variables in healthy children and adults
Meybosch S, et al.
Testing, 14(1), e0211212-e0211212 (2019)
In vitro tumorsphere formation assays
Johnson S, et al.
Bio-protocol, 3(3), e325-e325 (2013)
Cancer-associated fibroblasts induce epithelial-mesenchymal transition through secreted cytokines in endometrial cancer cells
Wang X, et al.
Oncology Letters, 15(4), 5694-5702 (2018)
Gab3 overexpression in human glioma mediates Akt activation and tumor cell proliferation
Jia P, et al.
Testing, 12(3), e0173473-e0173473 (2017)
A Dudley et al.
Oncogene, 33(41), 4952-4960 (2013-10-22)
Glioblastoma (GBM) is the most common and invasive adult brain cancer. The rapid invasion of cancer cells into the normal brain is a major cause of treatment failure, yet the mechanisms that regulate this process are poorly understood. We have

Articles

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

Discover answers to your frequently asked questions about animal component-free, xenobiotic-free, and chemically defined media and reagents for cell culture.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service