Skip to Content
Merck
All Photos(1)

Documents

H9380

Sigma-Aldrich

Sodium hippurate hydrate

≥99%

Synonym(s):

Benzoylaminoacetic acid sodium salt, Hippuric acid sodium salt hydrate, N-Benzoylglycine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CONHCH2COONa · xH2O
CAS Number:
Molecular Weight:
201.15 (anhydrous basis)
Beilstein:
3638470
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Sodium hippurate hydrate, ≥99%

Assay

≥99%

form

powder

color

white

application(s)

detection

SMILES string

O.[Na+].[O-]C(=O)CNC(=O)c1ccccc1

InChI

1S/C9H9NO3.Na.H2O/c11-8(12)6-10-9(13)7-4-2-1-3-5-7;;/h1-5H,6H2,(H,10,13)(H,11,12);;1H2/q;+1;/p-1

InChI key

IOZRWVOSUXHESF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hippuric acid is a low molecular weight phenolic carboxylic acid urinary metabolite that may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyu-Bong Kim et al.
Journal of toxicology and environmental health. Part A, 73(21-22), 1420-1430 (2010-10-19)
Toxicometabolomics of urinary biomarkers for human gastric cancer in a mouse model was investigated using (1)H-nuclear magnetic resonance (NMR) spectroscopy. A human gastric adenocarcinoma cell line (1 × 10(7) cells/ml) was grafted onto the skin of the back of intact
Luisa M Ostertag et al.
Molecular nutrition & food research, 55(11), 1624-1636 (2011-09-08)
Bioactive polyphenols from fruits, vegetables, and beverages have anti-platelet effects and may thus affect the development of cardiovascular disease. We screened the effects of 26 low molecular weight phenolic compounds on two in vitro measures of human platelet function. After
Jin Hee Kim et al.
Industrial health, 49(1), 8-14 (2010-09-09)
Exposure to volatile organic compounds (VOCs) was known to result in immunologic, respiratory, carcinogenic, reproductive, neurologic, and cardiovascular effects. However, the mechanisms by which VOCs induce these adverse health effects are not well understood. To evaluate the change of oxidative
Lindsey E Romick-Rosendale et al.
Arthritis research & therapy, 13(6), R199-R199 (2011-12-14)
Systemic lupus erythematosus (SLE or lupus) is a chronic autoimmune disease, and kidney involvement with SLE, a.k.a. lupus nephritis (LN), is a frequent and severe complication of SLE that increases patient morbidity and mortality. About 50% of patients with SLE
Kazuki Akira et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 35(4), 404-412 (2011-11-18)
Spontaneously hypertensive rats (SHR) provide a simple model for studying essential hypertension. Their genetic and metabolic features are of great interest because they may provide insights into the pathophysiological processes underlying essential hypertension. We have thus investigated the metabolic characteristics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service