Skip to Content
Merck
All Photos(3)

Key Documents

H7039

Sigma-Aldrich

3-Hydroxy-3-methylglutaryl-CoA reductase human

solution

Synonym(s):

HMGCoA Reductase, HMGR

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

Assay

≥90% (SDS-PAGE)

Quality Level

form

solution

specific activity

2-8 units/mg protein

mol wt

~76 kDa (for the fusion protein)

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HMGCR(3156)

Application

3-Hydroxy-3-methylglutaryl-CoA reductase human has been used:
  • in anti-HMGCR enzyme linked immunosorbent assay (ELISA)
  • as an antigen to measure anti-HMGCR antibodies by blot analysis
  • in HMG-CoA reductase activity assays
  • as an antigen to compare in-house antigen via sodium dodecyl sulphate-polyacrylamide gel electrophoresis (SDS-PAGE)/western blot

Biochem/physiol Actions

3-Hydroxy-3-methylglutaryl-CoA reductase/HMGCR serves as a rate limiting enzyme in mevalonate pathway. It also controls the oncogenic Hippo transducers TAZ/YAP. HMGCR also perform as a genetic modifier for Alzheimer′s disease (AD).
Hydroxy-3-methylglutaryl-CoA reductase catalyzes the committed step in cholesterol biosynthesis.

Physical properties

Consists of the HMG CoA reductase catalytic domain fused to GST protein.

Unit Definition

One unit will convert 1.0 μmole of NADPH to NADP+ per minute at 37 °C.

Physical form

Supplied as a solution containing 250 μg protein in 50 mM Tris pH 7.5, with 5 mM DTT, 1:200 Protease Inhibitor Cocktail (Catalog Number P8340) and 50% (w/v) glycerol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myopathy with anti-HMGCR antibodies Perimysium and myofiber pathology
Alshehri A, et al.
Neurology? neuroimmunology & neuroinflammation, 2(4), e124-e124 (2015)
HMG-CoAR expression in male breast cancer: relationship with hormone receptors, Hippo transducers and survival outcomes
Di BA, et al.
Scientific reports, 6, 35121-35121 (2016)
Marie Hudson et al.
Medicine, 95(44), e5280-e5280 (2016-11-20)
The objective of this study was to investigate the frequency of autoantibodies to hydroxymethylglutaryl coenzyme A reductase (HMGCR) in systemic sclerosis (SSc) and associations with inflammatory myositis and statin exposure.This was a cross-sectional, multicenter study of 306 subjects from the
ChemVassa: a new method for identifying small molecule hits in drug discovery
Moldover B, et al.
The Open Medicinal Chemistry Journal, 6, 29-29 (2012)
OP0009 Distinctive Immunofluorescence Pattern in Statin-Associated Autoimmune Myopathy with Anti-HMGCR Autoantibodies
Alvarado-Cardenas M, et al.
Annals of the Rheumatic Diseases, 75, 56-56 (2016)

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service