Skip to Content
Merck
All Photos(3)

Key Documents

H3760

Sigma-Aldrich

Hemoglobin from bovine blood

dried erythrocytes

Synonym(s):

Bovine hemoglobin, Hb, Methemoglobin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42030116
NACRES:
NA.61

biological source

bovine blood

Quality Level

form

dried erythrocytes

mol wt

Mr ~64500

solubility

0.6 M HCl: soluble 20 mg/mL

UniProt accession no.

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.

Biochem/physiol Actions

Oxygen transporter, NO scavenger
Oxygen transporter. The Fe2+/Fe3+ balance is a physiological indicator of blood oxygenation; deoxygenated hemoglobin accessorizes a feedback loop by reducing nitrite to NO, a vasodilator which enhances blood flow to oxygen-deprived tissues.

Quality

Methemoglobin and oxyhemoglobin content is not determined.

Caution

Since native hemoglobin is readily oxidized in air, these preparations may be predominantly methemoglobin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Van Phuc Nguyen et al.
Journal of biophotonics, 10(4), 577-588 (2016-05-03)
A variety of thermal therapeutic methods have been investigated to treat bladder tumors but often cause bowel injury and bladder wall perforation due to high treatment dosage and limited clinical margins. The objective of the current study is to develop
Glutamic acid reactivity in heated protein and protein-alginate mixtures.
C. G. Oates, d. A. Ledward, J. R. Mitchell and Hodgson.
International Journal of Food Science and Technology, 5, 477-483 (1987)
Michael Helton et al.
Biomedical optics express, 13(3), 1595-1608 (2022-04-14)
Measurements of time-resolved reflectance from a homogenous turbid medium can be employed to retrieve the absolute values of its optical transport coefficients. However, the uncertainty in the temporal shift of the experimentally determined instrument response function (IRF) with respect to
Byoungwook Bang et al.
Current protocols in pharmacology, 72, 5-5 (2016-03-21)
Animal models of experimentally induced inflammatory bowel disease (IBD) are useful for understanding more about the mechanistic basis of the disease, identifying new targets for therapeutic intervention, and testing novel therapeutics. This unit provides detailed protocols for five widely used
Michael Helton et al.
Biomedical optics express, 11(8), 4366-4378 (2020-09-15)
A heuristic method for estimating the reduced scattering coefficient (µs') of turbid media using time-resolved reflectance is presented. The technique requires measurements of the distributions of times-of-flight (DTOF) of photons arriving at two identical detection channels placed at unique distances

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service