Skip to Content
Merck
All Photos(1)

Key Documents

H0266

Sigma-Aldrich

Hemoglobin A2, Ferrous Stabilized human

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42030116
NACRES:
NA.61

biological source

human

Quality Level

Assay

97-100% (agarose gel electrophoresis)

form

lyophilized powder

technique(s)

immunofluorescence: suitable

suitability

suitable for electrophoresis and chromatography standard

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HBA2(3040)

General description

Hemoglobin A2 (HBA2) is mapped to human chromosome 16p13.3. HBA2 is a minor hemoglobin component and comprises of two α and δ chains.
Hemoglobin is the major component of red blood cells, and is responsible for their red color. Its normal concentration in erythrocytes is 34%. Hemoglobin is the most important respiratory protein of vertebrates by virtue of its ability to transport oxygen from the lungs to body tissues, and to facilitate the return transport of carbon dioxide.
has not been tested for functional equivalence against native preparations (unlyophilized ferrous hemoglobins).

Application

Hemoglobin A2 was used in the determination of fetal hemoglobin by time-resolved immunofluorometric assay.
Hemoglobin A2, Ferrous Stabilized human has been used as a reference in reversed-phase high-performance liquid chromatography for quantifying δ-globin elution profile. It has also been used as calibrator in isotope dilution mass spectrometry (IDMS) measurements.

Biochem/physiol Actions

Elevated levels of Hemoglobin A2 (HBA2) is observed in patients with β-thalassemia trait.

Packaging

Package size indicates the amount of hemoglobin as determined by the procedure of Drabkin, D.L., J. Biol. Chem., 164, 703 (1946).

Reconstitution

When reconstituted with buffer, gives >90% ferrous hemoglobin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elevated hemoglobin A2 as a marker for beta-thalassemia trait in pregnant women
Ou Z, et al.
The Tohoku Journal of Experimental Medicine, 223(3), 223-226 (2011)
Rapid and Sensitive Assessment of Globin Chains for Gene and Cell Therapy of Hemoglobinopathies
Loucari C, et al.
Human Gene Therapy Methods, 29(1), 60-74 (2018)
Saqib Hussain Ansari et al.
JPMA. The Journal of the Pakistan Medical Association, 62(12), 1314-1317 (2013-07-23)
To screen immediate family members of thalassaemia patients for carrier identification and counselling. The cross-sectional study was conducted at an urban thalassaemia treatment and prevention centre in Karachi, Pakistan, from January to December 2008, and involved 188 siblings of 100
Burçin Nalbantoğlu et al.
Pediatric hematology and oncology, 29(5), 472-478 (2012-08-08)
Iron deficiency (IDA) and beta thalassemia trait (TT) are the most common causes of hypochromia and microcytosis. Many indices have been defined to quickly discriminate these similar entities via parameters obtained from automated blood cell analyzers. However, studies in the
Determination of HbA2 by quantitative bottom-up proteomics and isotope dilution mass spectrometry
Arsene C, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 487, 318-324 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service