Skip to Content
Merck
All Photos(1)

Key Documents

H3757

Sigma-Aldrich

Hyaluronidase from bovine testes

Type VIII, lyophilized powder, 300-1,000 U/mg

Synonym(s):

Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine testis

Quality Level

conjugate

conjugate (Glucosaminoglycan)

type

Type VIII

form

lyophilized powder

specific activity

300-1,000 U/mg

mol wt

~55 kDa (four subunits of 14 kDa each)

UniProt accession no.

relevant disease(s)

cancer

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Hyaluronidase degrades hyaluronan and has been found to be inappropriately regulated during cancer progression.
These enzymes randomly cleave β-N-acetylhexosamine-[1→4] glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

Preparation Note

Prepared from aseptic filled solution of Type I-S.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kohtaro Miyazawa et al.
Histochemistry and cell biology, 133(1), 125-134 (2009-10-16)
Membranous epithelial cells (M cells) of the follicle-associated epithelium in Peyer's patches have a high capacity for transcytosis of several viruses and microorganisms. Here, we report that we have successfully established a bovine intestinal epithelial cell line (BIE cells) and
Chaemoon Lim et al.
Journal of cancer prevention, 27(2), 122-128 (2022-07-23)
Osteosarcoma is the most frequent primary malignant bone tumor with higher incidences in children and adolescents. Despite clinical evolutions, patients with osteosacoma have had a poor prognosis. There has been increasing evidence that cancer is a stem cell disease. This
Takayuki Yamochi et al.
Journal of assisted reproduction and genetics, 38(10), 2591-2599 (2021-05-11)
To enhance the in vitro growth of porcine oocytes, we studied the effect of mural granulosa cells (MGCs) on the viability of oocytes attached to granulosa cells (oocyte-granulosa cell complexes, OGCs) that were obtained from early antral follicles. When OGCs
C Rollo et al.
Reproduction (Cambridge, England), 154(4), 375-385 (2017-09-08)
Acetylation of histone proteins is a major determinant of chromatin structure and function. Fertilisation triggers a round of chromatin remodelling that prepares the genome for the first round of transcription from the new embryonic genome. In this study we confirm
Nathan V Welham et al.
Journal of speech, language, and hearing research : JSLHR, 52(4), 1008-1020 (2009-07-31)
To develop and evaluate a rat excised larynx model for the measurement of acoustic, aerodynamic, and vocal fold vibratory changes resulting from vocal fold scar. Twenty-four 4-month-old male Sprague-Dawley rats were assigned to 1 of 4 experimental groups: chronic vocal

Articles

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

See All

Protocols

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

Questions

1–4 of 4 Questions  
  1. What is the recommended method for reconstituting item H375, Hyaluronidase from bovine testes?

    1 answer
    1. The solubility of H375, Hyaluronidase from bovine testes, is 100 mg/mL in water. To achieve a higher solubility, it is advised to warm the tube at 37°C and shake it in an ultrasonic bath for a period. The stock solution can be stored at temperatures below -20°C for several months.

      Helpful?

  2. Good Morning, I our lab we got this product (H3757) and we would like to know how to obtain the package insert since we are not sure how to store it or after open expiration date. Not lot number on the bottle so not sure how to look for it. Thank you

    1 answer
    1. Hyaluronidase from bovine testes may be reconstituted at 1 mg/mL in 0.02 M sodium phosphate buffer, pH 7.0 containing 77mM sodium chloride and 0.01% bovine serum albumin. For more information please view the assay below:

      https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/enzyme-activity-assays/enzymatic-assay-of-hyaluronidase

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How is product H3757, Hyaluronidase from bovine testes, solubilized?

    1 answer
    1. Product H3757, Hyaluronidase from bovine testes, may be reconstituted at 1 mg/mL in 0.02 M sodium phosphate buffer, pH 7.0 containing 77mM sodium chloride and 0.01% bovine serum albumin.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service