Skip to Content
Merck
All Photos(1)

Key Documents

56690

Sigma-Aldrich

Hypericin from Hypericum perforatum

~95% (HPLC)

Synonym(s):

4,5,7,4′,5′,7′-Hexahydroxy-2,2′-dimethylnaphthodianthrone, Cyclo werrol, Hypericum red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H16O8
CAS Number:
Molecular Weight:
504.44
Beilstein:
1917913
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

~95% (HPLC)

Quality Level

solubility

1 M NaOH: 10 mg/mL, turbid, dark green

SMILES string

Cc1cc(O)c2C(=O)c3c(O)cc(O)c4c5c(O)cc(O)c6C(=O)c7c(O)cc(C)c8c1c2c(c34)c(c78)c56

InChI

1S/C30H16O8/c1-7-3-9(31)19-23-15(7)16-8(2)4-10(32)20-24(16)28-26-18(12(34)6-14(36)22(26)30(20)38)17-11(33)5-13(35)21(29(19)37)25(17)27(23)28/h3-6,31-36H,1-2H3

InChI key

BTXNYTINYBABQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hypericin hydrophobic is a red color pigment occurring naturally in Hypericum perforatum. Its fluorescence is measured at 600 nm.[1] It is localized in the intracellular membranes including nuclear membrane, mitochondrial membrane, Golgi apparatus, lysosomes and endoplasmic reticulum.[2]

Application

Hypericin from Hypericum perforatum has been used:
  • in the broth macrodilution test and antibacterial assay against bacterial strains[3]
  • as a medium supplement for human malignant melanoma cells for irradiation and localization studies[2]
  • as a ligand for pregnane X receptor (PXR) and as defatting medium supplement for primary human hepatocytes culture[4]

Biochem/physiol Actions

Anthraquinone-derivative within St. John′s wort. Antibacterial effective against Gram-positive and Gram-negative bacteria.
Hypericin is a potent inhibitor of protein kinase C (PKC), telomerase, cytochrome P450 and reverse transcriptase.[1] It is exploited for its photosensitizing functionality in photodynamic therapy (PDT) and photodynamic diagnosis (PDD) in cancer cells.[5]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Antibacterial effect of hypericin
Feyziouglu B, et al.
African Journal of Microbiology Research, 7(11), 979-982 (2013)
I Lopez-Bazzocchi et al.
Photochemistry and photobiology, 54(1), 95-98 (1991-07-01)
The polycyclic compound hypericin, a known photodynamic agent, was investigated for antiviral activity in the presence and absence of light. The three viruses tested: murine cytomegalovirus; Sindbis virus; and human immunodeficiency virus type 1, were all susceptible to hypericin; but
Susceptibility of In Vitro Melanoma Skin Cancer to Photoactivated Hypericin versus Aluminium (III) Phthalocyanine Chloride Tetrasulphonate
Ndhundhuma IM and Abrahamse H
BioMed Research International, 2017 (2017)
Hypericin in the light and in the dark: two sides of the same coin
Jendvzelovska Z, et al.
Frontiers in Plant Science, 7, 560-560 (2016)
Fleury Augustin Nsole Biteghe et al.
Journal of photochemistry and photobiology. B, Biology, 211, 111982-111982 (2020-09-01)
Aberrant anti-cancer drug efflux mediated by membrane protein ABC transporters (ABCB5 and ABCG2) is thought to characterize melanoma heterogeneous chemoresistant populations, presumed to have unlimited proliferative and self-renewal abilities. Therefore, this study primarily aimed to investigate whether continuous exposure of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service