Skip to Content
Merck
All Photos(1)

Key Documents

H1506

Sigma-Aldrich

3α-Hydroxysteroid Dehydrogenase from Pseudomonas testosteroni

≥15 units/mg protein

Synonym(s):

3α-Hydroxysteroid:NAD(P)+ oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Pseudomonas testosteroni)

form

lyophilized solid

specific activity

≥15 units/mg protein

purified by

chromatography

composition

Protein, ≥25%

foreign activity

β-hydroxysteroid dehydrogenase ≤0.5%
alcohol dehydrogenase ≤0.05%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

3α-Hydroxysteroid Dehydrogenase from Pseudomonas testosterone has been used:
  • in the preparation of composite electrode platform to for the determination of androsterone
  • in potassium phosphate buffer for the bioluminescent assay of bile acids in serum
  • for total bile salts assay using an enzymatic method

Biochem/physiol Actions

3α-hydroxysteroid dehydrogenase (3α-HSD) may help to modulate the occupancy of the androgen receptor (AR) in androgen target tissues. It may also act in hindering androgen action.

Unit Definition

One unit will oxidize 1.0 μmole of androsterone per min at pH 8.9 at 25 °C in the presence of β-NAD+.

Physical form

Lyophilized powder containing potassium phosphate buffer salt, EDTA, and DL-dithiothreitol

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Li et al.
Archives of pharmacal research, 30(7), 812-815 (2007-08-21)
Two novel lactones, Cephalosporolides H (1) and 1 (2), were isolated from the lyophilized culture broth of the marine-derived fungus, Penicillium sp. The structures were elucidated on the basis of extensive 1D- and 2D-NMR, as well as HRESI-MS spectroscopic analyses.
Bin Li et al.
Proteins, 71(2), 670-683 (2007-11-03)
Experimentally determined protein tertiary structures are rapidly accumulating in a database, partly due to the structural genomics projects. Included are proteins of unknown function, whose function has not been investigated by experiments and was not able to be predicted by
Laurence Meyer et al.
Neurobiology of disease, 30(1), 30-41 (2008-02-23)
Identification of cellular targets pertinent for the development of effective therapies against pathological pain constitutes a difficult challenge. We combined several approaches to show that 3alpha-hydroxysteroid oxido-reductase (3alpha-HSOR), abundantly expressed in the spinal cord (SC), is a key target, the
Enzyme biosensor for androsterone based on 3alpha-hydroxysteroid dehydrogenase immobilized onto a carbon nanotubes/ionic liquid/NAD+ composite electrode
Mundaca RA, et al.
Talanta, 99(, 697-702 (2012)
Bioluminescent assay for total bile acids in serum with use of bacterial luciferase.
Styrelius I, et al.
Clinical Chemistry, 29(6), 1123-1127 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service