Skip to Content
Merck
All Photos(6)

Documents

T0875

Sigma-Aldrich

Sodium taurodeoxycholate hydrate

≥95% (HPLC)

Synonym(s):

2-([3α,12α-Dihydroxy-24-oxo-5β-cholan-24-yl]amino)ethanesulfonic acid, Taurodeoxycholic acid sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C26H44NO6SNa · xH2O
CAS Number:
Molecular Weight:
521.69 (anhydrous basis)
Beilstein:
3901204
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Assay

≥95% (HPLC)

mol wt

micellar avg mol wt 3100

aggregation number

6

CMC

1-4 mM (20-25°C)

application(s)

sample preservation

SMILES string

O.[Na+].C[C@H](CCC(=O)NCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C26H45NO6S.Na.H2O/c1-16(4-9-24(30)27-12-13-34(31,32)33)20-7-8-21-19-6-5-17-14-18(28)10-11-25(17,2)22(19)15-23(29)26(20,21)3;;/h16-23,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);;1H2/q;+1;/p-1/t16-,17-,18-,19+,20-,21+,22+,23+,25+,26-;;/m1../s1

InChI key

OLPIZAYYAVQETM-GGPRKOIFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium taurodeoxycholate hydrate has been used in a study to assess the effect of submicellar concentrations of bile salts on the lipid bilayer membrane. It has also been used in a study to investigate polymorphic behavior in protein-surfactant mixtures.
Bile salt-related, anionic detergent used for isolation of membrane proteins including inner mitochondrial membrane proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Orioni et al.
The journal of physical chemistry. B, 110(24), 12129-12140 (2006-06-28)
Mixtures containing water, bovine serum albumin (BSA), and sodium taurodeoxycholate (NaTDC), a component of the bile in mammals, have been investigated in a wide range of composition and pH. Depending on the concentration of both solutes and the pH, solutions
Jing-Yi Ye et al.
International journal of nanomedicine, 15, 6503-6518 (2020-09-15)
A non-lipolysis nanoemulsion (NNE) was designed to reduce the first-pass metabolism of raloxifene (RAL) by intestinal UDP-glucuronosyltransferases (UGTs) for increasing the oral absorption of RAL, coupled with in vitro and in vivo studies. In vitro stability of NNE was evaluated
Monalisa Mohapatra et al.
Langmuir : the ACS journal of surfaces and colloids, 27(22), 13461-13467 (2011-10-07)
The interaction of submicellar concentrations of various physiologically important unconjugated [sodium deoxycholate (NaDC), sodium cholate (NaC)] and conjugated [sodium glycodeoxycholate (NaGDC), sodium glycocholate (NaGC), sodium taurodeoxycholate (NaTDC), sodium taurocholate (NaTC)] bile salts with dipalmitoylphosphatidylcholine (DPPC) and dimyristoylphosphatidylcholine (DMPC) small unilamellar
Nitya Subrahmanian et al.
Genetics, 214(4), 895-911 (2020-02-23)
Complex I is the first enzyme involved in the mitochondrial electron transport chain. With >40 subunits of dual genetic origin, the biogenesis of complex I is highly intricate and poorly understood. We used Chlamydomonas reinhardtii as a model system to
Xiao-Feng Zhang et al.
Food chemistry, 274, 305-313 (2018-10-31)
The low expression level of acidic lipases from Aspergillus sp. remains a major obstacle for their use in industrial applications. In this study, fusion expression with three fusion partners was investigated to enhance the expression level of an acidic lipase

Protocols

Investigate bile acid roles in gut hormone profiles and glycemic control, vital for clinical labs exploring potential mechanisms.

Investigate bile acid roles in gut hormone profiles and glycemic control, vital for clinical labs exploring potential mechanisms.

Investigate bile acid roles in gut hormone profiles and glycemic control, vital for clinical labs exploring potential mechanisms.

Investigate bile acid roles in gut hormone profiles and glycemic control, vital for clinical labs exploring potential mechanisms.

Related Content

Bile Acids (BA) are synthesized in the liver and play important roles in cholesterol homeostasis, absorption of vitamins and lipids, and various key metabolic processes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service