Skip to Content
Merck
All Photos(4)

Key Documents

H0501

Sigma-Aldrich

DL-Homocystine

≥98% (TLC)

Synonym(s):

meso-4,4′-Dithio-bis(2-aminobutanoic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(NH2)CH2CH2SSCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
268.35
Beilstein:
1728581
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
42021308
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-Homocystine,

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

>300 °C

application(s)

detection

SMILES string

N[C@H](CCSSCC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6+

InChI key

ZTVZLYBCZNMWCF-OLQVQODUSA-N

Gene Information

human ... BHMT(635)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Li et al.
Amino acids, 50(10), 1485-1494 (2018-08-01)
Plasma homocysteine is elevated in patients with impaired renal function, and markedly so at end-stage renal disease. As chronic kidney disease and hyperhomocysteinemia are also independent risk factors for cardiovascular disease, the latter is hypothesized to accelerate vascular abnormalities following
Sezgin Bakirdere et al.
Analytica chimica acta, 680(1-2), 41-47 (2010-10-26)
An analytical method was developed for the determination of thiols in biological samples. Reverse phase chromatography coupled to ICP quadrupole MS or Orbitrap MS was employed for the separation and detection of thiols. For the determination of total thiols, oxidized
Javier Escobar et al.
Journal of pharmaceutical and biomedical analysis, 123, 104-112 (2016-02-20)
The aminothiols are biological compounds with numerous vital functions. One of the most relevant is their role as antioxidant systems. The reduced to oxidized ratios are extremely useful indicators of oxidative stress and cellular redox status. We have validated an
M W Orth et al.
The Journal of nutrition, 122(3), 482-487 (1992-03-01)
The effect of excessive levels of cysteine and homocysteine on tibial dyschondroplasia (TD) in broiler chicks was studied. In the first experiment, graded levels of L-cysteine as well as one level of L-homocysteine were supplemented to a corn-soybean-based diet adequate
Chasity B Love-Nkansah et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(3), 934-940 (2015-11-05)
Homocysteine sulfinyl radical ((SO⋅) Hcy) is a reactive intermediate involved during oxidative damage of DNA in the presence of high concentrations of homocysteine (Hcy). The short lifetime of (SO⋅) Hcy makes its preparation, isolation, and characterization challenging using traditional chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service