Skip to Content
Merck
All Photos(1)

Documents

GERPN4301

AMDEX Goat Anti Rabbit IgG Horseradish Peroxidase Conjugate

Cytiva RPN4301

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

antibody product type

secondary antibodies

form

liquid

packaging

pkg of 0.5 mL

manufacturer/tradename

Cytiva RPN4301

storage temp.

2-8°C

General description

Goat Anti-Rabbit IgG - Horseradish Peroxidase. Goat anti-rabbit IgG conjugated to multiple HRP molecules on a Dextran backbone.
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein and mainly helps in immune defense. IgG is a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Rabbit IgG is further divided into five classes- IgM, IgG, IgA, IgD and IgE. It has only one IgG variant with one interheavy disulfide bridge. It helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Application

AMDEX goat anti rabbit IgG horseradish peroxidase conjugate has been used in immunohistochemistry and ELISA.

Features and Benefits

  • It can be used in direct substitution for a standard anti-rabbit HRP conjugate, without the need to alter existing protocols.
  • It is important to optimize the dilution by setting up a range test.
  • Option to reduce incubation times.
  • Choice of six novel reagents.
  • Well-controlled nonspecific binding properties.

Storage and Stability

Please be aware this product may be shipped 30 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

AMDEX is a trademark of Cytiva

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular Genetics of Immunoglobulin (1987)
Emily N Kirby et al.
Innate immunity, 30(2-4), 40-54 (2024-01-23)
Interferon Stimulated Gene (ISG) expression plays a key role in the control of viral replication and development of a robust adaptive response. Understanding this dynamic relationship between the pathogen and host is critical to our understanding of viral life-cycles and
Highly efficient detection of salbutamol in environmental water samples by an enzyme immunoassay.
Liu Z, et al.
The Science of the Total Environment, 613, 861-865 (2018)
The Immunoglobulins: Structure and Function (1998)
The Laboratory Rat (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service