GE29401323
StrepTrap™ XT prepacked chromatography column
pack of 5 × 5 mL, prepacked chromatography column
Synonym(s):
StrepTrap™ High Performance
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106501
NACRES:
SB.53
Recommended Products
grade
for chromatography
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 29401323
Looking for similar products? Visit Product Comparison Guide
General description
StrepTrap XT, a ready-to-use prepacked chromatography column, is packed with Strep-Tactin XT Sepharose resin for protein purification.
Features and Benefits
- Achieve highly pure Strep-tag II and Twin-Strep-tag recombinant proteins
- Experience fast purifications with simple elution
- Physiological conditions and mild elution preserve target protein activity
- Ensures quick and easy regeneration
- Compatible with a wide range of detergents, denaturants, and other additives
Legal Information
StrepTrap is a trademark of Cytiva
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service