Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
StrepTrap
™
XT prepacked chromatography column
Synonym(s):
StrepTrap
™
High Performance
Products
Product Category
columns (4)
Brand
Sigma-Aldrich (4)
Manufacturer
Physical Form
Separation Technique
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GE29401323
StrepTrap
™
XT prepacked chromatography column,
pack of 5 × 5 mL, prepacked chromatography column
Expand
View Pricing
GE29401322
StrepTrap
™
XT prepacked chromatography column,
pkg of 1 × 5 mL, prepacked chromatography column
Expand
View Pricing
GE29401320
StrepTrap
™
XT prepacked chromatography column,
pack of 5 × 1 mL, prepacked chromatography column
Expand
View Pricing
GE29401317
StrepTrap
™
XT prepacked chromatography column,
pkg of 1 × 1 mL, prepacked chromatography column
Expand
View Pricing