GE28-9031-32
Protein A High Performance Spintrap™
Cytiva 28-9031-32, pack of 16 columns
Synonym(s):
High Performance Spintrap, Protein A Spintrap
About This Item
Recommended Products
ligand
native protein A
feature
wetted part: no
packaging
pack of 16 columns
manufacturer/tradename
Cytiva 28-9031-32
storage condition
(20% Ethanol)
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
cleaning
2-10
working range
3-9
capacity
>1 mg binding capacity(human IgG well)
storage temp.
2-8°C
General description
Application
There is also a protocol for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min. No pretreatment of the sample is required.
Features and Benefits
- For simple, small-scale antibody purification or enrichment of specific Proteins in prepacked, single-use spin columns that are ready to use.
- Classic and cross-link protocols provide flexibility.
- Elution conditions formatted for both electrophoresis and LC-MS analysis workflows.
- Fast binding kinetics and high capacity provide high yield.
- Prepacked with Protein A Sepharose™ High Performance for coupling of antibodies of IgG subclasses.
- Easy scale-up with HiTrap® Protein A HP prepacked columns.
Storage and Stability
Analysis Note
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page shows various purification options for Protein A Sepharose chromatography media and describes typical binding and elution conditions for Protein A Sepharose chromatography media.
This page describes immunoprecipitation (immunoaffinity or pull-down techniques).
Protocols
This page shows how to separate IgG antibodies by affinity chromatography using Protein A HP SpinTrap columns from Cytiva.
This page shows how to separate IgG antibodies by affinity chromatography using Protein G HP SpinTrap™/Ab SpinTrap™ from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service