Skip to Content
Merck
All Photos(1)

Key Documents

GAGO20

Sigma-Aldrich

Glucose (GO) Assay Kit

sufficient for 20 assays

Synonym(s):

Glucose Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84
Pricing and availability is not currently available.

usage

sufficient for 20 assays

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

This kit is for the quantitative, enzymatic determination of glucose in food and other materials. Glucose is oxidized to gluconic acid and hydrogen peroxide by glucose oxidase. Hydrogen peroxide reacts with o-dianisidine in the presence of peroxidase to form a colored product. Oxidized o-dianisidine reacts with sulfuric acid to form a more stable colored product. The intensity of the pink color measured at 540 nm is proportional to the original glucose concentration.

Suitability

Suitable for the quantitative, enzymatic determination of glucosein food and other materials.

Kit Components Only

Product No.
Description

  • o-dianisidine reagent 1 mL/vial

  • glucose oxidase/peroxidase reagent 1 capsule

  • glucose standard .5 mL

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bergmeyer, H.U. and Bernt, E.,
Methods of Enzymatic Analysis, 1205-1212 (1974)
S Talukder et al.
Journal of food science and technology, 57(2), 537-548 (2020-03-03)
Natural plant pigment, anthocyanins have the capability to change its color with the change of its structure influenced by changing pH. This feature of anthocyanin has been harnessed to design a meat products quality indicator. In the present experiment anthocyanin
M Isabel Muro-Pastor et al.
Plant physiology, 184(4), 1792-1810 (2020-09-10)
Cyanobacteria unable to fix atmospheric nitrogen have evolved sophisticated adaptations to survive to long periods of nitrogen starvation. These genetic programs are still largely unknown-as evidenced by the many proteins whose expression is regulated in response to nitrogen availability, but
Nelson H Knudsen et al.
Science (New York, N.Y.), 368(6490) (2020-05-02)
Repeated bouts of exercise condition muscle mitochondria to meet increased energy demand-an adaptive response associated with improved metabolic fitness. We found that the type 2 cytokine interleukin-13 (IL-13) is induced in exercising muscle, where it orchestrates metabolic reprogramming that preserves
David Ohayon et al.
Nature materials, 19(4), 456-463 (2019-12-18)
A promising class of materials for applications that rely on electron transfer for signal generation are the n-type semiconducting polymers. Here we demonstrate the integration of an n-type conjugated polymer with a redox enzyme for the autonomous detection of glucose

Questions

1–10 of 15 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Why would liquid samples need to be diluted to 20-80 ug/mL? I'm hoping to be able to use this kit to measure glucose levels in serum that should be between 5 - 30 mmol/L. Would this work?

    1 answer
    1. Unfortunately, no, this kit is not suitable for concentrations of 5-30 mmol/L, which equates to 900-5400 mg/L. This concentration is far above the detection range and would offer poor results. The detection range of this kit is approximately 10-100 ug/ml and sample dilution is required for accurate, repeatable results. Measuring samples outside of this range is not recommended.

      Helpful?

  4. Can I use this kit for colorimetric tests?

    1 answer
    1. Yes, this kit is suitable for use with any colorimeter capable of measuring absorbance at 540 nm. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/516/298/gago20bul.pdf

      Helpful?

  5. Is it possible to adapt the GAGO20 Glucose (GO) Assay Kit to a 96-well plate?

    1 answer
    1. Researchers have documented an adaptation of the kit to a 96-well assay format as follows.
      To prepare glucose standards, 16 µl of 1 mg/ml glucose should be diluted with 84 µl PBS to achieve a 100 µl final volume for a 0.16 mg/ml standard. Four 2-fold serial dilutions into PBS (50 µl 0.16 mg/ml + 50 µl PBS for 0.08 mg/ml standard, etc.) should be done to generate 0.01, 0.02, 0.04, 0.08, and 0.16 mg/ml glucose standards. These assays are linear from 0.01 to 0.16 mg/ml glucose. Glucose stock solutions can be stored at −20°C or 4°C.
      Then, 30 µl of glucose standards and a PBS blank should be added to the first row of a clear-bottom 96 well plate. In the next row, 30 µl of each sample should be added into an individual well after diluting them 1:4–1:8 with PBS.

      For Glucose Oxidase-based Glucose assay:
      • Add 100 µl of GO reagent (Sigma; GAGO-20) to each well with a multichannel pipette.
      • Seal the wells with parafilm to prevent evaporation and incubate the plate at 37°C for 30–60 min.
      • Stop the reaction by adding 100 µl of 12 N H2SO4 (the samples should visibly change color from yellow/orange to pink).
      • Centrifuge the plate in an appropriate swing-bucket rotor to clear condensate from the sides of the wells and to remove any air bubbles present in the samples.
      • Use a plate reader to measure absorbance at 540 nm.
      • Determine the free glucose concentration by comparing the free glucose measurements for each sample to the glucose standard curve.

      Helpful?

  6. Can I scale down the reaction volumes 20-25 times lower than what is specified in the protocol? So instead of using 1ml of sample, 2 ml of assay reagent, and 2ml of sulfuric acid, use 50ul, 100ul and 100ul of each step and react?

    1 answer
    1. This kit has been optimized for use at the volumes given in the protocol. The suitability of using smaller volumes in a tube format would have to be determined by the end user and is not recommended. However, other researchers have published an adaptation of the kit to a 96-well assay format which utilizes similar volumes. Please note that this process has not been validated. The 96-well plate protocol can be reviewed below.

      Prepare glucose standards: Dilute 16 µl of 1 mg/ml glucose with 84 µl PBS (100 µl final volume) for 0.16 mg/ml standard. Do four 2-fold serial dilutions into PBS (50 µl 0.16 mg/ml + 50 µl PBS for 0.08 mg/ml standard, etc.) to generate 0.01, 0.02, 0.04, 0.08, and 0.16 mg/ml glucose standards. These assays are linear from 0.01 to 0.16 mg/ml glucose. Glucose stock solutions can be stored at −20°C or 4°C.

      Add 30 µl of glucose standards and a PBS blank to the first row of a clear-bottom 96 well plate. In the next row, add 30 µl of each sample into an individual well (after diluting them 1:4–1:8 with PBS).
      9a)

      For Glucose Oxidase-based Glucose assay:
      Add 100 µl of GO reagent (Sigma; GAGO-20) to each well with a multichannel pipette. Seal the wells with parafilm to prevent evaporation and incubate the plate at 37°C for 30–60 min. Stop the reaction by adding 100 µl of 12 N H2SO4 (the samples should visibly change color from yellow/orange to pink). Centrifuge the plate in an appropriate swing-bucket rotor to clear condensate from the sides of the wells and to remove any air bubbles present in the samples. Use a plate reader to measure absorbance at 540 nm. Determine the free glucose concentration by comparing the free glucose measurements for each sample to the glucose standard curve.

      Please see this article for more information: https://www.ncbi.nlm.nih.gov/pmc/articles/PMC4048761/

      Helpful?

  7. Can I perform the reaction on polystyrene based plastic tube? ie do the reaction directly on 96-well clear plate

    1 answer
    1. Researchers have published an adaptation of the kit to a 96-well assay format as follows. Please note that this process has not been validated.

      Prepare glucose standards: Dilute 16 µl of 1 mg/ml glucose with 84 µl PBS (100 µl final volume) for 0.16 mg/ml standard. Do four 2-fold serial dilutions into PBS (50 µl 0.16 mg/ml + 50 µl PBS for 0.08 mg/ml standard, etc.) to generate 0.01, 0.02, 0.04, 0.08, and 0.16 mg/ml glucose standards. These assays are linear from 0.01 to 0.16 mg/ml glucose. Glucose stock solutions can be stored at −20°C or 4°C.

      Add 30 µl of glucose standards and a PBS blank to the first row of a clear-bottom 96 well plate. In the next row, add 30 µl of each sample into an individual well (after diluting them 1:4–1:8 with PBS).
      9a)

      For Glucose Oxidase-based Glucose assay:
      Add 100 µl of GO reagent (Sigma; GAGO-20) to each well with a multichannel pipette. Seal the wells with parafilm to prevent evaporation and incubate the plate at 37°C for 30–60 min. Stop the reaction by adding 100 µl of 12 N H2SO4 (the samples should visibly change color from yellow/orange to pink). Centrifuge the plate in an appropriate swing-bucket rotor to clear condensate from the sides of the wells and remove any air bubbles present in the samples. Use a plate reader to measure absorbance at 540 nm. Determine the free glucose concentration by comparing the free glucose measurements for each sample to the glucose standard curve.

      Please see this article for more information: https://www.ncbi.nlm.nih.gov/pmc/articles/PMC4048761/

      Helpful?

  8. How would you deproteinate and decolorize serum sample to be compatible with the GAGO20 assay kit?

    1 answer
    1. There are a number of ways to deproteinize serum. Most classical methods use acids or solvents to precipitate proteins followed by centrifugation. One example is given below:
      Use perchloric acid (10% v/v) in a volume equal to the sample, mix well, and centrifuge. Remove the supernatant and neutralize it with KOH (20% w/v). Allow to stand for 10 minutes in an ice bath and centrifuge. The sample (supernatant) can then be run in the assay.

      Another option is filtration through a 10 kDa MWCO spin filter. The spin filter that can be used for this purpose is SKU UFC5010.
      https://www.sigmaaldrich.com/US/en/product/mm/ufc5010

      Note that high hemoglobin levels (caused by hemolysis upon collection) may also contribute to the colorization of serum and plasma, however upon dilution, it should not be a factor with GAGO20.

      See the link below to a publication that reviews other methods of deproteinization:
      https://www.longdom.org/open-access/deproteination-of-blood-plasma-in-human-body-for-serum-analysis-93551.html

      Helpful?

  9. Can Product GAGO20, Glucose (GO) Assay Kit, be used in a 96-well plate format?

    1 answer
    1. Sigma Aldrich has not performed this in a 96-well plate format.  However if a method has have not tried this. If you have developed a method in a 96-well format, Sigma Aldrich would be happy to know about this.

      Helpful?

  10. What is the difference between the GAGO20 and GAHK20 Glucose (GO) Assay Kits?

    1 answer
    1. The kits use different enzymes. GAGO20 uses glucose oxidase and peroxidase, while GAHK20 uses hexokinase.  Furthermore, the final reaction products are monitored at different wavelengths, 540 nm (GAGO20) and 340 nm (GAHK20).

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service