Skip to Content
Merck
All Photos(1)

Key Documents

G3660

Supelco

Glucose oxidase/peroxidase reagent

for use with enzymatic assay kits GAGO20, STA20

Synonym(s):

GOD-POD Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

Quality Level

analyte chemical class(es)

sugars (glucose)

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

Glucose oxidase/peroxidase reagent along with O-Dianisidine was used for the determination of blood and urinary glucose, especially the presence of fructose for both research and clinical purpose. It may be used for determining glucose in plasma or serum. It may also be used in the estimation of blood glucose as a function of the oxidative coupling system or of the oxygen affinity of the oxygen acceptor chromogen.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Single glucose oxidase-peroxidase reagent for two-minute determination of serum glucose.
L G Morin et al.
Clinical chemistry, 19(9), 959-962 (1973-09-01)
Interference in glucose oxidase-peroxidase blood glucose methods.
P Sharp
Clinica chimica acta; international journal of clinical chemistry, 40(1), 115-120 (1972-08-01)
Ketty Margulis et al.
Life (Basel, Switzerland), 10(9) (2020-09-10)
Pgr5 proteins play a major direct role in cyclic electron flow paths in plants and eukaryotic phytoplankton. The genomes of many cyanobacterial species code for Pgr5-like proteins but their function is still uncertain. Here, we present evidence that supports a
Use of glucose oxidase, peroxidase, and O-dianisidine in determination of blood and urinary glucose.
Huggett AG and Nixon DA.
Lancet, 270 (6991), 368-370 (1957)
Takashi Nishimura
Current biology : CB, 30(18), 3624-3632 (2020-07-18)
Metabolism must be coupled with developmental transition to fulfill the energy requirements during an organism's life cycle. In most animals, steroid hormones are crucial regulators of life-stage transitions until adulthood. In the fruit fly Drosophila, ecdysteroid titers drive developmental transitions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service