Skip to Content
Merck
All Photos(1)

Key Documents

G7646

Sigma-Aldrich

β-Glucuronidase from Escherichia coli

Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

type

Type VII-A

Quality Level

form

lyophilized powder

specific activity

5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)

mol wt

290 kDa

composition

Protein, ~25% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
Effective in the hydrolysis of steroid glucuronides.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Quality

Contains buffer salts and stabilizer.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at the pH 6.8 (30 min assay).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimized conditions for the emzymatic hydrolysis of a-hydroxytriazolam-glucoronide in human urine.
Tsujikawa, K., et al.
Health Laboratory Science, 40, 286-289 (2004)
H Mita et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 34(8), 1262-1269 (2004-08-10)
Aspirin challenge of aspirin-intolerant asthma (AIA) patients causes a significant increase in leukotriene E4 (LTE4) concentration in urine. However, knowledge on leukotriene B4 (LTB4) generation in patients with AIA is insufficient. Recent research has demonstrated that exogenously administered LTB4 is
Lawrence D Joh et al.
Biotechnology progress, 22(3), 723-730 (2006-06-03)
Extraction and storage of a recombinant protein produced by transient expression following agroinfiltration of lettuce were investigated. Lettuce leaves expressing beta-glucuronidase (GUS) were extracted by homogenization in several buffer combinations, and the yield and stability were assessed. The reducing agent
M Coulet et al.
Journal of veterinary pharmacology and therapeutics, 25(2), 89-97 (2002-05-10)
The pharmacokinetics of ibafloxacin, a new veterinary fluoroquinolone antimicrobial agent, was studied following intravenous (i.v.) and oral administration to healthy dogs. The mean absolute bioavailability of ibafloxacin after oral doses of 7.5, 15 and 30 mg/kg ranged from 69 to
Enzymes
Hans Ulrich Bergmeyer
Methods of Enzymatic Analysis, 460-461 (1974)

Articles

β-glucuronidase (GUS) enzymes are utilized to hydrolyze glucuronide (gluc) drug metabolites to the parent drug, facilitating analysis by LC-MS/MS.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

See All

Protocols

Objective: To standardize a procedure for the enzymatic assay of β-Glucuronidase.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

See All

Questions

1–2 of 2 Questions  
  1. It is recommended to resuspend the product in which buffer?

    1 answer
    1. This enzyme is lyophilized from a solution containing buffer salts and a stabilizer. The product may be reconsituted at 1 mg/mL in water. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/412/974/g7646dat.pdf

      Helpful?

  2. Is BSA or other protein components included in the overall percentage listed on the CoA for the product G7646, which is β-Glucuronidase from Escherichia coli (CAS 9001-45-0)?

    1 answer
    1. No. BSA or other protein sources are added to the product except Potassium Phosphate and Sucrose. Although this is a fermentation product, there may be other protein residues in it in a negligible amount. The product is tested by activity, residual protein contaminants are not tested. The protein percentage is the total protein (tested by Biuret method).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service