Skip to Content
Merck
All Photos(6)

Key Documents

G6129

Sigma-Aldrich

Guanosine 3′,5′-cyclic monophosphate sodium salt

≥99% (HPLC), powder

Synonym(s):

3′,5′-Cyclic GMP monosodium salt, 3′,5′-cGMP-Na, Guanosine 3′,5′-cyclophosphate monosodium salt, cGMP, cyclic GMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5NaO7P
CAS Number:
Molecular Weight:
367.19
Beilstein:
6467425
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

powder

color

white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O)C(=O)N1

InChI

1S/C10H12N5O7P.Na/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6;/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17);/q;+1/p-1/t3-,5-,6-,9-;/m1./s1

InChI key

KMPIYXNEROUNOG-GWTDSMLYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

An important second messenger, cGMP is a major intracellular mediator of extracellular signals such as nitric oxide and natriuretic peptides. Cyclic GMP interacts with three types of intracellular receptor proteins: cGMP-dependent protein kinases, cGMP-regulated channels, and cGMP-regulated cyclic nucleotide phosphodiesterases. A primary action of elevated cGMP levels in vivo is the stimulation of cGMP-dependent protein kinase (PKG).

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases and PKA & PKG pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Picco et al.
The Journal of physiology, 460, 741-758 (1993-01-01)
1. The permeability of the channel activated by guanosine 3',5'-cyclic monophosphate (cGMP) to many organic monovalent cations was determined by recording macroscopic currents in excised inside-out patches of plasma membrane from isolated retinal rod outer segments of the tiger salamander.
G Colamartino et al.
The Journal of physiology, 440, 189-206 (1991-01-01)
1. Blockage and permeation of divalent cations through channels activated by guanosine 3',5'-cyclic monophosphate (cyclic GMP) were studied in membrane patches excised from retinal rods of the tiger salamander Ambystoma tigrinum by rapidly changing the ionic medium bathing the intracellular
Milton Packer et al.
Circulation, 131(1), 54-61 (2014-11-19)
Clinical trials in heart failure have focused on the improvement in symptoms or decreases in the risk of death and other cardiovascular events. Little is known about the effect of drugs on the risk of clinical deterioration in surviving patients.
Brandon A Kemp et al.
Circulation research, 115(3), 388-399 (2014-06-07)
Compound 21 (C-21) is a highly selective nonpeptide AT2 receptor (AT2R) agonist. To test the hypothesis that renal proximal tubule AT2Rs induce natriuresis and lower blood pressure in Sprague-Dawley rats and mice. In rats, AT2R activation with intravenous C-21 increased
Sofia-Iris Bibli et al.
Cardiovascular research, 106(3), 432-442 (2015-04-15)
H2S is known to confer cardioprotection; however, the pathways mediating its effects in vivo remain incompletely understood. The purpose of the present study is to evaluate the contribution of cGMP-regulated pathways in the infarct-limiting effect of H2S in vivo. Anaesthetized

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service