Skip to Content
Merck
All Photos(2)

Key Documents

G5419

Sigma-Aldrich

Anti-GAD1/GAD2 Antibody

mouse monoclonal, K-87

Synonym(s):

Anti-GAD 67

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-Glutamic Acid Decarboxylase 67 (GAD67) antibody produced in mouse, clone K-87, purified antibody

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

K-87, monoclonal

mol wt

antigen 67 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500-1:20,000 (with antigen-retrieval step)
western blot: 1:500-1:20,000 using mouse brain extract

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... GAD1(2571)
mouse ... Gad1(14415)
rat ... Gad1(24379)

General description

Glutamic Acid Decarboxylase 67 (GAD67) is encoded by the gene glutamic acid decarboxylase 1 (GAD1). GAD1 gene is located on the human chromosome at 2q31.1. GAD67 is localized in the dendrites and cell bodies.

Specificity

Reacts specifically with GAD67. Does not crossreact with GAD65.

Immunogen

This peptide does not share homology with GAD65.
synthetic peptide from mouse GAD67 (amino acids 87-106).

Application

Monoclonal Anti-Glutamic Acid Decarboxylase 67 (GAD67) antibody is suitable for use in immunoblot, western blot (at 1:500-1:20,000 dilutions using mouse brain extract) and immunohistochemistry (at 1:500-1:20,000 dilutions in mouse brain sections).
Monoclonal anti-glutamic acid decarboxylase 67 (GAD67) antibody produced in mouse has been used in:
  • immunohistofluorescence
  • western blotting
  • immunohistochemistry.

Biochem/physiol Actions

Glutamic Acid Decarboxylase 67 (GAD67) is involved in the production of γ-amino butyric acid (GABA), a inhibitory neurotransmitter in the central nervous system (CNS). Deficiency of GAD67 leads to psychosis, schizophrenia and bipolar diseases.

Physical form

Solution in phosphate buffered saline containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjung Kim et al.
PloS one, 7(2), e32043-e32043 (2012-03-01)
The analysis of co-localized protein expression in a tissue section is often conducted with immunofluorescence histochemical staining which is typically visualized in localized regions. On the other hand, chromogenic immunohistochemical staining, in general, is not suitable for the detection of
Ying Chen et al.
Neuropharmacology, 60(7-8), 1075-1087 (2010-09-28)
GAD67 corresponds to one of two enzymes that decarboxylates glutamate to produce γ-aminobutyric acid, the main inhibitory neurotransmitter in the mammalian central nervous system, hence defining the cellular phenotype of a diverse set of inhibitory interneurons of the brain. Reduced
A Guidotti et al.
Archives of general psychiatry, 57(11), 1061-1069 (2000-11-14)
Reelin (RELN) is a glycoprotein secreted preferentially by cortical gamma-aminobutyric acid-ergic (GABAergic) interneurons (layers I and II) that binds to integrin receptors located on dendritic spines of pyramidal neurons or on GABAergic interneurons of layers III through V expressing the
Silvia Rossi et al.
Brain, behavior, and immunity, 25(5), 947-956 (2010-10-14)
Synaptic dysfunction triggers neuronal damage in experimental autoimmune encephalomyelitis (EAE), a model of multiple sclerosis (MS). While excessive glutamate signaling has been reported in the striatum of EAE, it is still uncertain whether GABA synapses are altered. Electrophysiological recordings showed
Baohu Ji et al.
PloS one, 8(6), e66327-e66327 (2013-07-05)
It has been well established that schizophrenia patients display impaired NMDA receptor (NMDAR) functions as well as exacerbation of symptoms in response to NMDAR antagonists. Abnormal NMDAR signaling presumably contributes to cognitive deficits which substantially contribute to functional disability in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service