Skip to Content
Merck
All Photos(1)

Key Documents

G1774

Sigma-Aldrich

Glucagon

≥95% (HPLC), powder, synthetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C153H225N43O49S
CAS Number:
Molecular Weight:
3482.75
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32
biological source:
synthetic
form:
powder
Assay:
≥95% (HPLC)
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

sterility

non-sterile

Assay

≥95% (HPLC)

form

powder

technique(s)

cell based assay: suitable

solubility

1% acetic acid: 1.00-1.04 mg/mL, clear, colorless
water: 1.00-1.04 mg/mL, clear, colorless

suitability

suitable for molecular biology

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl[H].[H]N[C@@H](Cc1cnc[nH]1)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)NCC(=O)N[C@@H](C(C)O)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H](C(C)O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc5ccccc5)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc6c[nH]c7ccccc67)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](C(C)O)C(O)=O

InChI

1S/C153H225N43O49S.ClH/c1-72(2)52-97(133(226)176-96(47-51-246-11)132(225)184-104(60-115(159)209)143(236)196-123(78(10)203)151(244)245)179-137(230)103(58-83-64-167-89-29-19-18-28-87(83)89)183-131(224)95(43-46-114(158)208)177-148(241)120(74(5)6)194-141(234)101(54-79-24-14-12-15-25-79)182-138(231)105(61-117(211)212)185-130(223)94(42-45-113(157)207)171-124(217)75(7)170-127(220)91(31-22-49-165-152(160)161)172-128(221)92(32-23-50-166-153(162)163)174-146(239)110(69-199)191-140(233)107(63-119(215)216)186-134(227)98(53-73(3)4)178-135(228)99(56-81-33-37-85(204)38-34-81)180-129(222)90(30-20-21-48-154)173-145(238)109(68-198)190-136(229)100(57-82-35-39-86(205)40-36-82)181-139(232)106(62-118(213)214)187-147(240)111(70-200)192-150(243)122(77(9)202)195-142(235)102(55-80-26-16-13-17-27-80)188-149(242)121(76(8)201)193-116(210)66-168-126(219)93(41-44-112(156)206)175-144(237)108(67-197)189-125(218)88(155)59-84-65-164-71-169-84;/h12-19,24-29,33-40,64-65,71-78,88,90-111,120-123,167,197-205H,20-23,30-32,41-63,66-70,154-155H2,1-11H3,(H2,156,206)(H2,157,207)(H2,158,208)(H2,159,209)(H,164,169)(H,168,219)(H,170,220)(H,171,217)(H,172,221)(H,173,238)(H,174,239)(H,175,237)(H,176,226)(H,177,241)(H,178,228)(H,179,230)(H,180,222)(H,181,232)(H,182,231)(H,183,224)(H,184,225)(H,185,223)(H,186,227)(H,187,240)(H,188,242)(H,189,218)(H,190,229)(H,191,233)(H,192,243)(H,193,210)(H,194,234)(H,195,235)(H,196,236)(H,211,212)(H,213,214)(H,215,216)(H,244,245)(H4,160,161,165)(H4,162,163,166);1H/t75-,76+,77+,78+,88-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,120-,121-,122-,123-;/m0./s1

InChI key

RKGLLHCSSVJTAN-YYICOITRSA-N

Gene Information

human ... GCG(2641)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr

General description

Glucagon (GCG) peptide is secreted in response to insulin from pancreatic islets α-cells.[1]
Glucagon is synthesized from proglucagon and is 29-amino acid peptide hormone.[2] Proglucagon gene is mapped to human chromosome 2q24.2.[3]

Application

Glucagon (GCG) has been:
  • used as a component of hormone stock solution for preserving full biological activity of heart tissues obtained from male Sprague-Dawley rats[1]
  • used as an infusion in phases II and III fasting king penguins to study the various lipolytic, metabolic, and hormonal responses[4]
  • used for the stimulation of PGC-1α expression in hepatocytes[5]
  • used to induce the expression of methionine adenosyltransferase α1 (MAT1A), which is involved in the regulation of hepatic levels of S-adenosylmethionine and the adaptive response to fasting[6]
  • used to study its effect on stimulation of gluconeogenesis through hepatic lipolysis, mediated by inositol trisphosphate receptor 1 (INSP3R1)[7]
  • intraperitoneally injected in mice to assess glucagon-induced Sam68 subcellular localization in vivo[8]

Biochem/physiol Actions

Glucagon (GCG) produces elevation in contractility and the frequency of contraction in heart muscles. It activates PI3K (phosphoinositide 3-kinase) and thus, elevates glucose utilization by heart muscles.[1] In multiple mammals, though not humans, this peptide induces lipolysis, and is the major lipolytic hormone in birds. It also induces gluconeogenesis and glycogenolysis in mammals and birds.[4]
The paired box 6 (PAX6) regulates the expression of glucagon.[9] High levels of glucagon referred to as hyperglucagonemia leads to a high risk of diabetes and is associated with pancreatic neuroendocrine tumors.[2] Defective glucagon hypersecretion is regarded as an etiopathogenic factor.[9]

Components

H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH.xHCl

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucagon stimulates gluconeogenesis by INSP3R1-mediated hepatic lipolysis
Perry RJ, et al.
Nature, 579(7798), 279-283 (2020)
Glucose-dependent downregulation of glucagon gene expression mediated by selective interactions between ALX3 and PAX6 in mouse alpha cells
Mirasierra M and Vallejo M
Diabetologia, 59(4), 766-775 (2016)
Baicalin ameliorates hepatic insulin resistance and gluconeogenic activity through inhibition of p38 MAPK/PGC-1alpha pathway
Fang P, et al.
Phytomedicine, 64, 153074-153074 (2019)
Association of TCF7L2 and GCG gene variants with insulin secretion, insulin resistance, and obesity in new-onset diabetes
Zhang L, et al.
Biomedical and Environmental Sciences : Bes, 29(11), 814-817 (2016)
Aijun Qiao et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Hepatic glucose production (HGP) is an important component of glucose homeostasis, and deregulated HGP, particularly through gluconeogenesis, contributes to hyperglycemia and pathology of type-2 diabetes (T2D). It has been shown that the gluconeogenic gene expression is governed primarily by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service