Skip to Content
Merck
All Photos(1)

Documents

G2501

Sigma-Aldrich

L-Glutamic Dehydrogenase from bovine liver

Type I, ammonium sulfate suspension, ≥40 units/mg protein

Synonym(s):

L-GLDH, L-Glutamate:NAD[P]+ Oxidoreductase (deaminating), Glutamate Dehydrogenase from bovine liver

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type I

form

ammonium sulfate suspension

specific activity

≥40 units/mg protein

mol wt

310-350 kDa

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-glutamic dehydrogenase catalyzes the conversion of glutamate to α-ketoglutarate.
Mammalian forms of this enzyme, including this bovine form, can use either NADP(H) or NAD(H) as coenzymes. L-glutamic dehydrogenase plays a unique role in mammalian metabolism. The reverse reaction catalyzed by this enzyme is the only pathway by which ammonia can become bound to the α-carbon atom of an α-carboxylic acid and thus, is the only source of de novo amino acid synthesis in mammalian species.

The bovine enzyme is characterized by three sets of properties:
  • It has a reversible concentration-dependent association, producing higher molecular weight forms.
  • Forms tight enzyme-reduced coenzyme-substrate ternary complexes whose rates of dissociation modulate the steady-state reaction rates.
  • Exhibits a wide variety of effects from the binding of any of a number of nucleotide modifiers.

L-glutamic dehydrogenase catalyzes the conversion of glutamate to α-ketoglutarate.

Unit Definition

One unit will reduce 1.0 μmole of α-ketoglutarate to L-glutamate per min at pH 7.3 at 25 °C, in the presence of ammonium ions.

Physical form

Suspension in 2.0 M (NH4)2SO4 solution

Analysis Note

Protein determined by biuret

substrate

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehran Karimi et al.
Seminars in thrombosis and hemostasis, 35(4), 426-438 (2009-07-15)
Factor XIII (FXIII) is a tetrameric zymogen (FXIII-A (2)B (2)) that is converted into an active transglutaminase (FXIIIa) by thrombin and Ca (2+) in the terminal phase of the clotting cascade. By cross-linking fibrin chains and alpha (2) plasmin inhibitor
Hyperinsulinism and hyperammonaemia syndrome and severe myoclonic epilepsy of infancy.
F Pérez Errazquin et al.
Neurologia (Barcelona, Spain), 26(4), 248-252 (2010-12-18)
Shanti Balasubramaniam et al.
Journal of pediatric endocrinology & metabolism : JPEM, 24(7-8), 573-577 (2011-09-22)
Hyperinsulinism-hyperammonemia syndrome (HI/HA) (OMIM 606762), the second most common form of congenital hyperinsulinism (CHI) is associated with activating missense mutations in the GLUD1 gene, which encodes the mitochondrial matrix enzyme, glutamate dehydrogenase (GDH). Patients present with recurrent symptomatic postprandial hypoglycemia
Cleanthe Spanaki et al.
Neurotoxicity research, 21(1), 117-127 (2011-11-01)
Glutamate dehydrogenase (GDH) catalyzes the reversible inter-conversion of glutamate to α-ketoglutarate and ammonia. High levels of GDH activity is found in mammalian liver, kidney, brain, and pancreas. In the liver, GDH reaction appears to be close-to-equilibrium, providing the appropriate ratio
Michisuke Yuzaki
The European journal of neuroscience, 32(2), 191-197 (2010-07-22)
Several C1q family members, especially the Cbln and C1q-like subfamilies, are highly and predominantly expressed in the central nervous system. Cbln1, a member of the Cbln subfamily, plays two unique roles at parallel fiber (PF)-Purkinje cell synapses in the cerebellum:

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service