Skip to Content
Merck
All Photos(3)

Key Documents

G1135

Sigma-Aldrich

L-Glutamic acid γ-(4-nitroanilide)

γ-glutamyl transpeptidase substrate

Synonym(s):

L-γ-Glutamyl-p-nitroanilide, L-Glutamic acid 5-(4-nitroanilide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H13N3O5
CAS Number:
Molecular Weight:
285.25
Beilstein:
2818758
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥98% (HPLC)

form

powder

solubility

formic acid: 50 mg/mL, clear to slightly hazy

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)Nc1ccc(cc1)[N+]([O-])=O)C(O)=O

InChI

1S/C11H13N3O5/c12-9(11(16)17)5-6-10(15)13-7-1-3-8(4-2-7)14(18)19/h1-4,9H,5-6,12H2,(H,13,15)(H,16,17)/t9-/m0/s1

InChI key

WMZTYIRRBCGARG-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Substrates

Substrate for γ-glutamyl transpeptidase

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Dvoráková et al.
General physiology and biophysics, 15(5), 403-413 (1996-10-01)
The initial rate kinetics of rat kidney gamma-glutamyl transpeptidase were measured using L-gamma-glutamyl-p-nitroanilide and glycyl-glycine as the donor and the acceptor substrate, respectively. Experimental data were fitted with the initial rate equation, and the obtained results indicated that: (1) Michaelis
Gololobov MYu et al.
The Biochemical journal, 304 ( Pt 3), 869-876 (1994-12-15)
Acyl-transfer catalysed by gamma-glutamyltranspeptidase from bovine kidney was studied using gamma-L- and gamma-D-Glu-p-nitroanilide as the donor and GlyGly as the acceptor. The transfer of the gamma-Glu group to GlyGly was shown to be accompanied by transfer of the gamma-Glu group
Man Tong et al.
Cell reports, 36(8), 109617-109617 (2021-08-26)
The liver plays central roles in coordinating different metabolic processes, such as the catabolism of amino acids. In this study, we identify a loss of tyrosine catabolism and a concomitant increase in serum tyrosine levels during liver cancer development. Liver
M Moriguchi et al.
Archives of microbiology, 144(1), 15-19 (1986-02-01)
Three gamma-glutamyltranspeptidase (enzymes I, II and III) were partially purified from the cell free extracts of the cultured mycelia of Morchella esculenta Fr. The molecular masses of enzymes were 155,000 (I), 219,000 (II) and 102,000 (III). All of them catalyzed
P M Verhoeff et al.
Clinica chimica acta; international journal of clinical chemistry, 175(2), 129-134 (1988-07-15)
In this paper we compare the measurement of catalytic activity concentrations of gamma-glutamyltransferase with the non-carboxylated and the carboxylated substrate in preparations of different origin. Fresh human sera, commercial test sera and preparations of gamma-glutamyltransferase purified from human liver, porcine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service