Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.84
eCl@ss:
32160414
UNSPSC Code:
41116133
form
lyophilized powder
Quality Level
usage
vial sufficient for ≥50 assays
technique(s)
PCR: suitable
storage temp.
−20°C
General description
The Adenosine 5’-triphosphate (ATP) Bioluminescent Assay Kit may be employed for the quantitative bioluminescent determination of adenosine 5’-triphosphate (ATP) in samples containing 2x10–12 to 8x10–5 moles/liter. ATP is consumed and light is emitted when luciferase catalyzes the oxidation of D-luciferin.
Application
The ATP assay kit has been used to measure the level of cellular ATP present in isolated mitochondrial fractions from L. donovani promastigotes.
Adenosine 5′-triphosphate (ATP) assay mix has also been used to measure the level of cellular ATP in ATP viability assay.
Preparation Note
Reconstitute with 5 ml of sterile water.
Other Notes
Contains luciferase, luciferin, MgSO4, EDTA, DTT, BSA, and Tricine buffer.
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Guiping Sui et al.
American journal of physiology. Renal physiology, 306(3), F286-F298 (2013-11-29)
The urothelium is a newly recognized sensory structure that detects bladder fullness. Pivotal to this sensory role is the release of ATP from the urothelium. However, the routes for urothelial ATP release, its modulation by receptor-mediated pathways, and the autocrine/paracrine
Buse Cevatemre et al.
Chemico-biological interactions, 260, 248-255 (2016-10-11)
Despite the rapid advances in chemotherapy regimens, the outcome of patients with breast cancer is not satisfactory. One of the reasons of this dissatisfaction is that subsets of cells in tumors which referred as cancer stem cells (CSCs) show and/or
C J Williams et al.
Osteoarthritis and cartilage, 26(6), 797-806 (2018-03-27)
Mutations on chromosomes 5p (CCAL2) and 8q (CCAL1) have been linked to familial forms of calcium pyrophosphate deposition disease (CPDD). Mutations in the ANKH gene account for CCAL2, but the identity of CCAL1 has been elusive. Recently, a single Dutch
Global Trade Item Number
| SKU | GTIN |
|---|---|
| FLAAM-1VL | 04061833622865 |
| FLAAM-5VL | 04061833622872 |
| FLAAM-10VL | 04061833622841 |
