Skip to Content
Merck
All Photos(1)

Key Documents

F5386

Sigma-Aldrich

Fibrin from human plasma

insoluble powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human plasma

form

powder

technique(s)

indirect ELISA: suitable

UniProt accession no.

SMILES string

N(CC(=O)NC)C(=O)CN

InChI

1S/C5H11N3O2/c1-7-5(10)3-8-4(9)2-6/h2-3,6H2,1H3,(H,7,10)(H,8,9)

InChI key

BWGVNKXGVNDBDI-UHFFFAOYSA-N

Gene Information

General description

Fibrin is produced from cross-linking of fibrinogen by thrombin. Fibrinogen is made up of two monomers each containing three polypeptide chains. Fibrinogen is a major plasma protein.

Application

Fibrin from human plasma has been used:
  • for in vitro cleavage of fibrinogen and fibrin
  • as a reference for true component analysis (TCA) and principal component analysis (PCA) of Raman spectra
  • as a reference in Raman spectroscopy to study the binding mechanisms and the binding efficiency of the different polyoxometalates (POMs)

Biochem/physiol Actions

Fibrin acts as an important bio-polymer in the primary blood clotting process. It creates a scaffold for infiltrating cells during tissue repair. Fibrin is widely used as a tissue sealant for tissue regeneration applications as it enables the formation of a biocompatible physical structure. It is also used in delivery systems for cells and bioactive molecules. The fibrin matrix resembles the native pancreas and can serve as a scaffold material for islet transplantation. Fibrin has been used in studies to obtain fibrin-specific monoclonal antibodies (mAbs). These mAbs may be used to determine their role in fibrin polymerization and use in fibrin quantification in blood plasma.

Preparation Note

Prepared by clotting fibrinogen to yield a crosslinked fibrin

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of etanercept on lung leukocyte margination and fibrin deposition after cardiac surgery.
Barry Dixon et al.
American journal of respiratory and critical care medicine, 188(6), 751-754 (2013-09-17)
Suk Ho Bhang et al.
Biomaterials, 34(30), 7247-7256 (2013-07-06)
While subcutaneous tissue has been proposed as a potential site for pancreatic islet transplantation, concern remains that the microvasculature of subcutaneous tissue is too poor to support transplanted islets. In an effort to overcome this limitation, we evaluated whether fibrin
Craig P Fischer et al.
Journal of the American College of Surgeons, 217(3), 385-393 (2013-08-24)
This study evaluated the hemostatic effectiveness and safety of Fibrin Pad (Omrix Biopharmaceuticals Ltd.) vs absorbable hemostat in patients undergoing nonemergent surgery. Fibrin Pad is a topical absorbable hemostat designed to be effective in a variety of soft tissues and
Matteo Centola et al.
Tissue engineering. Part A, 19(17-18), 1960-1971 (2013-04-25)
Standard cartilage tissue engineering approaches, for example, matrix-induced autologous chondrocyte implantation (MACI), consist of the implantation of cell-based constructs whose survival and further development first depend on the degree of graft maturity at the time of surgery (e.g., matrix production)
Kristen T Morin et al.
Experimental cell research, 319(16), 2409-2417 (2013-06-27)
In vitro models of endothelial assembly into microvessels are useful for the study of angiogenesis and vasculogenesis. In addition, such models may be used to provide the microvasculature required to sustain engineered tissues. A large range of in vitro models

Articles

Cell based angiogenesis assays to analyze new blood vessel formation for applications of cancer research, tissue regeneration and vascular biology.

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service