Skip to Content
Merck
All Photos(1)

Documents

F5135

Sigma-Aldrich

N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala

collagenase substrate, chromogenic

Synonym(s):

FALGPA, N-[3-(2-Furyl)acryloyl]-L-leucyl-glycyl-L-prolyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N4O7
CAS Number:
Molecular Weight:
476.52
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product name

N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala,

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)\C=C\c1ccco1)C(=O)NCC(=O)N2CCC[C@H]2C(=O)N[C@@H](C)C(O)=O

InChI

1S/C23H32N4O7/c1-14(2)12-17(26-19(28)9-8-16-6-5-11-34-16)21(30)24-13-20(29)27-10-4-7-18(27)22(31)25-15(3)23(32)33/h5-6,8-9,11,14-15,17-18H,4,7,10,12-13H2,1-3H3,(H,24,30)(H,25,31)(H,26,28)(H,32,33)/b9-8+/t15-,17-,18-/m0/s1

InChI key

ZLFQNOJSYZSINX-PVJKAEHXSA-N

Gene Information

mouse ... Prkcq(18761)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

FA-Leu-Gly-Pro-Ala

General description

N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala or FALGPA is a synthetic substance which resemble the primary structure of collagen, and is hydrolyzed by all known collagenases.

Application

N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala has been used for use in FALGPA assay performed using GBS (Group B Streptococci) USF704.

Biochem/physiol Actions

N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala or FALGPA is hydrolyzed by collagenases and the optimum pH for hydrolysis is 7.4.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Substrates

Substrate for collagenase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Söderling et al.
Journal of periodontal research, 26(1), 17-23 (1991-01-01)
Four rough-surfaced (R) and three smooth-surfaced (S) clinical isolates of Capnocytophaga obtained from the subgingival plaque of periodontitis patients were studied for their peptidase and protease profiles. The results were compared with those obtained with C. gingivalis (which has a
Z E Juarez et al.
Infection and immunity, 67(1), 271-278 (1998-12-24)
Streptococcus gordonii is a frequent cause of infective bacterial endocarditis, but its mechanisms of virulence are not well defined. In this study, streptococcal proteases were recovered from spent chemically defined medium (CDM) and fractionated by ammonium sulfate precipitation and by
M S Yu et al.
Microbiology (Reading, England), 145 ( Pt 1), 143-150 (1999-04-17)
The prtV gene, encoding a collagenase of Vibrio parahaemolyticus, was expressed in Escherichia coli and purified by affinity chromatography. The transformant E. coli BL21(DE3)(pPRT2) secreted the recombinant PrtV, and the highest enzyme activity was detected in the culture supernatant after
K K Mäkinen et al.
Medical microbiology and immunology, 185(1), 1-10 (1996-05-01)
Relatively scant chemical information has been available on the proteinases and peptidases of spirochetes in spite of the association of spirochetes with several serious infections known to plague humans and other animal species. This situation has partly resulted from difficulties
R Gayatri et al.
Biochimica et biophysica acta, 1524(2-3), 228-237 (2000-12-13)
Bacterial collagenase has now been reacted with a select series of Cr(III) complexes and modifications in the activity of chromium-modified collagenase has been deduced from the extent of hydrolysis of (2-furanacryloyl-L-leucyl-glycyl-L-prolyl-L-alanine), FALGPA. A homologous series of Cr(III) complexes with dimeric

Protocols

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

To measure collagenase activity, N-(3-[2-Furyl]acryloyl)-Leu-Gly-Pro-Ala is used in a continuous spectrophotometric rate determination at 345 nm. Collagenase hydrolyzes collagen peptide bonds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service