Skip to Content
Merck
All Photos(1)

Key Documents

F3040

Sigma-Aldrich

Monoclonal ANTI-FLAG® M1 antibody

clone M1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.32

biological source

(Bioreactor)

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M1, monoclonal

form

buffered aqueous solution

purified by

using Protein A

species reactivity

all

concentration

2-5 mg/mL

technique(s)

western blot: 10 μg/mL

immunogen sequence

DYKDDDDK

shipped in

dry ice

storage temp.

−20°C

General description

The ANTI-FLAG M1 IgG2b mooclonal antibody binds to proteins with a FLAG marker at the free N-terminus.

Method of purification - Protein A

Immunogen

FLAG; peptide sequence DYKDDDDK

Application

Monoclonal ANTI-FLAG® M1 antibody is suitable for immunoprecipitation, western blotting, or EIA in yeast, animal and E.coli cells.

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.


Chromatin immunoprecipitation (1 paper)


Immunofluorescence (1 paper)

Learn more product details in our FLAG® applications portal.

Biochem/physiol Actions

Binds to the FLAG epitope when it is located at the free amino-terminus of a fusion protein. Does not bind to Met-FLAG fusion proteins, so will not recognize unprocessed, cytoplasmically expressed proteins. Binding is Ca2+-dependent; the complex dissociates in the absence of calcium ions.

Physical form

Solution in 10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide (w/v).

Preparation Note

Dilute antibody to 10ug/mL in .05 M TBS, pH 7.4, containing 1 mM CaCl2.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Best et al.
Nature communications, 5, 4760-4760 (2014-09-12)
Alternative splicing--the production of multiple messenger RNA isoforms from a single gene--is regulated in part by RNA binding proteins. While the RBPs transformer2 alpha (Tra2α) and Tra2β have both been implicated in the regulation of alternative splicing, their relative contributions
Mohammad Al Sorkhy et al.
BMC cancer, 12, 45-45 (2012-01-28)
Spy1 is a novel 'cyclin-like' activator of the G1/S transition capable of enhancing cell proliferation as well as inhibiting apoptosis. Spy1 protein levels are tightly regulated during normal mammary development and forced overexpression in mammary mouse models accelerates mammary tumorigenesis.
Reem Berro et al.
Journal of virology, 82(14), 7155-7166 (2008-05-16)
The human immunodeficiency virus type 1 (HIV-1) Tat is a 14-kDa viral protein that acts as a potent transactivator by binding to the transactivation-responsive region, a structured RNA element located at the 5' end of all HIV-1 transcripts. Tat transactivates
Eric J Gapud et al.
Journal of immunology (Baltimore, Md. : 1950), 187(4), 1826-1834 (2011-07-12)
Ataxia telangiectasia mutated (ATM) and DNA-dependent protein kinase catalytic subunits (DNA-PKcs) are members of the phosphatidylinositol 3-like family of serine/threonine kinases that phosphorylate serines or threonines when positioned adjacent to a glutamine residue (SQ/TQ). Both kinases are activated rapidly by
Min Luo et al.
Cell research, 20(2), 211-222 (2010-01-27)
As a critical apoptosis executioner, caspase-3 becomes activated and then enters into the nucleus to exert its function. However, the molecular mechanism of this nuclear entry of active caspase-3 is still unknown. In this study, we revealed that caspase-3 harbors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service