Skip to Content
Merck
All Photos(3)

Key Documents

EC11303

Sigma-Aldrich

Escherichia coli

Strain B, lyophilized cells

Synonym(s):

E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

Quality Level

sterility

non-sterile (not processed or packaged aseptically)

form

lyophilized cells

suitability

not intended for starter culture

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Escherichia coli is a non-pathogenic, facultative anaerobic flora present in the intestine of human. It belongs to the genus, Escherichia, which consists of motile gram-negative bacilli.

Application

Escherichia coli has been used:
  • in the isolation of DNA from lyophilized cell
  • in the preparation of crude cell lysate for developing scalable purification method for recombinant protein A
  • as test proteome for comparative electrophoresis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Pacheco et al.
Mikrochimica acta, 189(5), 194-194 (2022-04-16)
Janus micromotors encapsulating transition metal dichalcogenides (TMDs) and modified with a rhodamine (RhO)-labeled affinity peptide (RhO-NFMESLPRLGMH) are used here for Salmonella enterica endotoxin detection. The OFF-ON strategy relies on the specific binding of the peptide with the TMDs to induce
Diarrheagenic Escherichia coli
Nataro JP and Kaper JB
Clinical Microbiology Reviews, 11(1), 142-201 (1998)
Horizontal comparative fluorescence two-dimensional gel electrophoresis for improved spot coordinate detection
Hanneken M, et al.
Electrophoresis, 35(8), 1118-1121 (2014)
A D Leung et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 38(5), 789-793 (2008-03-18)
About 90% of patients with atopic dermatitis (AD) are colonized with Staphylococcus aureus. S. aureus worsens AD by secreting superantigens and structural molecules within the cell wall that induce skin inflammation. Therefore, S. aureus in the home may contribute to
Severe atopic dermatitis is associated with a high burden of environmental Staphylococcus aureus
Leung AD, et al.
Clinical and Experimental Allergy, 38(5), 789-793 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service