Skip to Content
Merck
All Photos(1)

Key Documents

E9762

Sigma-Aldrich

Endoglycosidase F1 from Elizabethkingia miricola

recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution

Synonym(s):

Endo F1, Endo-β-N-acetylglucosaminidase F1, Endoglycosidase F1 from Chryseobacterium meningosepticum, Endoglycosidase F1 from Elizabethkingia meningoseptica, Endoglycosidase F1 from Flavobacterium meningosepticum

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

form

buffered aqueous solution

specific activity

≥16 U/mg

mol wt

32 kDa

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Endoglycosidase F1 from Elizabethkingia miricola is a glycan specific enzyme.

Application

Cleaves asparagine-linked or free oligomannose and hybrid, but not complex, oligosaccharides.
Endoglycosidase F1 from Elizabethkingia miricola has been used to remove binding of human hemochromatosis protein (HFE) to cation independent mannose-6-phosphate receptor (CI-MPR) and to glycosylate fucosylated N-glycopeptides.

Biochem/physiol Actions

Endoglycosidase F1 from Elizabethkingia miricola plays an important role in cleavage of glycan structures from the protein by cleaving between the two N-acetylglucosamine residues of the chitobiose core. It mediates high mannose and hybrid oligosaccharides cleavage.

Packaging

Supplied with 5× Reaction Buffer, 250 mM NaH2PO4, pH 5.5.

Unit Definition

One unit will release N-linked oligosaccharides from 1 μmole of denatured Ribonuclease B in 1 minute at 37 °C, pH 5.5.

Physical form

Aseptically filled solution in 20 mM Tris-HCl pH 7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Receptor activity-modifying protein 1 determines the species selectivity of non-peptide CGRP receptor antagonists
Mallee JJ, et al.
The Journal of Biological Chemistry, 277, 14294-14298 (2002)
EndoS2 is a unique and conserved enzyme of serotype M49 group A Streptococcus that hydrolyses N-linked glycans on IgG and alpha-acid glycoprotein
Jonathan S, et al.
BioChemistry: An Indian Journal, 455, 107-118 (2013)
Quantitative analysis of core fucosylation of serum proteins in liver diseases by LC-MS-MRM
Ma J, et al.
Journal of proteomics, 189, 67-74 (2018)
In vitro binding of HFE to the cation-independent mannose-6 phosphate receptor
Schimanski LM, et al.
Blood Cells, Molecules and Diseases, 43(2), 180-193 (2009)
Helena Ryšlavá et al.
The FEBS journal, 278(14), 2469-2484 (2011-05-14)
Fungal β-N-acetylhexosaminidases are inducible extracellular enzymes with many biotechnological applications. The enzyme from Penicillium oxalicum has unique enzymatic properties despite its close evolutionary relationship with other fungal hexosaminidases. It has high GalNAcase activity, tolerates substrates with the modified N-acyl group

Articles

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service