Skip to Content
Merck
All Photos(1)

Key Documents

324727

Sigma-Aldrich

Endoglycosidase F3, Elizabethkingia meningosepticum, Recombinant, E. coli

Endoglycosidase F3, Elizabethkingia meningosepticum, Recombinant, E. coli, cleaves asparagine-linked or free biantennary and triantennary complex, and Man3GlcNAc oligosaccharides from glycoproteins.

Synonym(s):

Endo-β-N-acetylglucosaminidase F3, Endo F3

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

conjugate

(N-linked)

form

liquid

specific activity

≥30 units/mg protein
≥5 units/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

Proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Recombinant, Elizabethkingia meningosepticum endoglycosidase F3 expressed in E. coli. Cleaves asparagine-linked or free biantennary and triantennary complex, and Man3GlcNAc oligosaccharides from glycoproteins. This enzyme cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine residue. Core fucosylation increases the activity of Endo F3 up to 40 fold. Exhibits no activity on high mannose and hybrid molecules. Less sensitive to protein conformation than N-Glycosidase F (Cat. No. 362185) and therefore is more suitable for deglycosylation of native proteins.
Recombinant, Elizabethkingia meningosepticum endoglycosidase F3 expressed in E. coli. Cleaves asparagine-linked or free biantennary and triantennary complex, and Man3GlcNAc oligosaccharides from glycoproteins. This enzyme cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide, generating a truncated sugar molecule with one N-acetylglucosamine residue remaining on the asparagine residue. Core fucosylation increases the activity of Endo F3 up to 40 fold. Exhibits no activity on high mannose and hybrid molecules. Less sensitive to protein conformation than N-Glycosidase F (Cat. No. 362185) and therefore is more suitable for deglycosylation of native proteins.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will release N-linked oligosaccharides from 1.0 µmol porcine fibrinogen per min at 37°C, pH 5.5.

Other Notes

Tarentino, A.L., et al. 1995. Glycobiology 5, 599.
Tarentino, A.L., and Plummer, T.H. 1994. Methods Enzymol. 230, 44.
Tarentino, A.L., et al. 1993. J. Biol. Chem. 268, 9702.
Trimble, R.B., and Tarentino, A.L. 1991. J. Biol. Chem. 266, 1646.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Tarentino et al.
Glycobiology, 5(6), 599-601 (1995-09-01)
The gene for endo-beta-N-acetylglucosaminidase F3 was cloned into the high-expression vector pMAL c-2, and expressed in Escherichia coli as a fusion protein. A key step in the purification employed Poros II (HS) chromatography, which greatly facilitated isolation of the enzyme
A L Tarentino et al.
The Journal of biological chemistry, 268(13), 9702-9708 (1993-05-05)
The genes for Flavobacterium meningosepticum Endo (endoglycosidase) F2 and Endo F3 were cloned, and their nucleotide sequences were determined. The deduced amino acid sequences were verified independently to a large extent by direct peptide microsequencing of 66 and 84% of
R B Trimble et al.
The Journal of biological chemistry, 266(3), 1646-1651 (1991-01-25)
Flavobacterium meningosepticum endo-beta-acetyl-glucosaminidase F preparations have been resolved by hydrophobic interaction chromatography on TSK-butyl resin into at least three activities designated endo F1, endo F2 and endo F3 each with a unique substrate specificity. The 32-kDa endo F1 protein is
Enzymatic deglycosylation of asparagine-linked glycans: purification, properties, and specificity of oligosaccharide-cleaving enzymes from Flavobacterium meningosepticum.
A L Tarentino et al.
Methods in enzymology, 230, 44-57 (1994-01-01)

Articles

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Explore strategies for releasing N-linked glycans with PNGase F, PNGase A & native & sequential deglycosylation with endoglycosidases & exoglycosidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service