Skip to Content
Merck
All Photos(1)

Key Documents

E8905

SAFC

Erythropoietin from rat

recombinant, expressed in insect cells, suitable for cell culture

Synonym(s):

EPO

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.75

recombinant

expressed in insect cells

mol wt

22-26 kDa by SDS-PAGE
calculated mol wt 18.5 kDa

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

−20°C

Biochem/physiol Actions

Erythropoietin, a glycoprotein produced primarily by the kidneys and at lower levels by the liver, is the primary regulatory factor of erythropoiesis. EPO promotes the proliferation, differentiation, and survival of the erythroid progenitors. EPO stimulates erythropoiesis by inducing growth and differentiation of burst forming units and colony forming units into mature red blood cells. EPO produced by kidney cells is increased in response to hypoxia or anemia.

The biological effects of erythropoietin are mediated by the erythropoietin receptor, which binds EPO with high affinity and is a potent EPO antagonist. When EPO is present at low concentrations, the EPO receptor initiates prolongation of G1 in the cell cycle and sends a differentiation signal; whereas at high EPO concentrations, a proliferation signal is generated and the G1 is shortened.
Erythropoietin is a glycoprotein that is the principal regulator of red blood cell growth and differentiation.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.5 mg bovine serum albumin.

Preparation Note

This Erythropoietin (EPO) is produced from a DNA sequence encoding rat erythropoietin (Met 1-Arg 192). Mature soluble recombinant rat EPO has Ala 27 at the amino terminus. The 166 amino acid residue soluble EPO has a calculated molecular mass of approximately 18.5 kDa. Due to glycosylation, the recombinant protein migrates as a 22-26 kDa in SDS-PAGE. Erythropoietin has been cloned from various species including human, murine, canine, etc. The mature proteins from the various species are highly conserved, exhibiting greater than 80% amino acid sequence identity. Rat EPO cDNA encodes a 192 amino acid residue precursor protein that is processed to yield a 166 amino acid residue mature protein. EPO contains 3 N-linked glycosylation sites. Glycosylation of EPO is required for the biological activities of EPO in vivo.

Analysis Note

The biological acitivity is measured by its ability to stimulate cell proliferation using TF-1 cell line.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagao, M., et al.
Biochimica et Biophysica Acta, 117, 99-99 (1992)
Ismayil Ahmet et al.
Biology open, 1(10), 1049-1053 (2012-12-06)
Activation of nitric oxide (NO) signaling is considered, at list partially, a mechanistic basis for EPO-induced cardioprotection. Surprisingly, hemodynamic response subsequent to NO activation after EPO administration has never been reported. The objectives of this study were to evaluate the
Yueyue Yu et al.
PloS one, 8(7), e69620-e69620 (2013-08-13)
Neonatal necrotizing enterocolitis (NEC) is a devastating gastrointestinal disease of preterm infants. Increased intestinal epithelium permeability is an early event in NEC pathogenesis. Autophagy and apoptosis are induced by multiple stress pathways which may impact the intestinal barrier, and they
Luca Weltert et al.
Transfusion, 55(7), 1644-1654 (2015-02-24)
We conducted a prospective single-blind randomized study to assess whether a single 80,000 IU dose of human recombinant erythropoietin (HRE), given just 2 days before cardiac surgery, could be effective in reducing perioperative allogeneic red blood cell transfusion (aRBCt). Six-hundred
Karin Lindahl et al.
Therapeutic drug monitoring, 37(6), 745-750 (2015-03-27)
The aim of the study was to investigate whether patients with a previous nonresponse to standard of care treatment with ribavirin dosed according to body weight would respond to a high individualized dose of concentration-monitored ribavirin. Previous nonresponders to standard

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service