Skip to Content
Merck

Skip To

E5546

Erythropoietin (EPO) human

recombinant, expressed in HEK 293 cells, suitable for cell culture

Synonym(s):

EPO, epoetin, hematopoietin, hemopoietin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Form:
lyophilized powder
Assay:
≥98% (SDS-PAGE)
Recombinant:
expressed in HEK 293 cells
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in HEK 293 cells

Quality Segment

assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≤2.5 ng/mL ED50 (measured in a cell proliferation assay using TF-1 human cells)

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

storage temp.

−20°C

General description

Recombinant human erythropoietin (EPO) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of ~18 kDa. This protein is manufactured in human cells using an all-human production system, with full chemically defined ingredients and with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports better stability of the protein in culture. The protein is produced with no artificial tags.

Biochem/physiol Actions

Erythropoietin (EPO) is a glycoprotein hormone, known for its erythropoietic activity. It is used as treatment for anemia in humans with renal failure or cancer. EPO also reduces the amount of blood transfusions required on premature infants and surgeries.
EPO consists of 165 amino acids and has a high carbohydrate content, of both N-linked and O-linked types. The oligosaccharide chains are responsible for EPO production, secretion, longevity, and functioning. The biological activity of EPO depends upon two disulfide bonds, one formed between cysteine7 and cysteine160, and the other between cysteine29 and cysteine33. Thus, EPO proper folding, glycosylation and disulfide bonds formation are crucial for its function.
EPO was found to downregulate HIF-1α in the diabetic rat model. EPO helped maintaining the homeostasis of intracellular zinc in retinal cells, altered as a result of diabetes, by restoring Zinc transporter 8 (ZnT8) expression.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H5166SRP6158E9530
assay

≥98% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥95% (SDS-PAGE)

assay

≥90% (SDS-PAGE)

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in HEK 293 cells

recombinant

expressed in NSO cells

form

lyophilized powder

form

lyophilized powder

form

lyophilized

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

impurities

Endotoxin, tested

impurities

≤1 EU/μg

impurities

-

impurities

endotoxin, tested


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the the total unit of E5546-50UG?

    1 answer
    1. The total unit of E5546-50UG is not provided as the product is characterized in micrograms (ug).

      Helpful?

Reviews

No rating value

Active Filters