Skip to Content
Merck
All Photos(2)

Key Documents

E8886

Sigma-Aldrich

Erythrosin extra bluish

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
Colour Index Number:
45430
Beilstein:
1443945
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Erythrosin extra bluish, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥80%

pH

6.4 (30.3 °C)

mp

315 °C

solubility

water: 1 mg/mL, clear

density

0.625-0.731 g/cm3 at 30.1 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5ccccc45)c1I

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;;/h1-6,25-26H;;/q;2*+1/p-2

InChI key

RAGZEDHHTPQLAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Erythrosin extra bluish is soluble in ethanol and water. It is a brown powder, with a pH range of 2.5 to 4. It is used in color filters, light emitting diodes, cosmetics and hair dyes.[1] It has been used in many biological applications such as, detecting gene expression, treating age related macular degeneration, diabetes and obesity.[2]

Suitability

Certified for use in the Jackson stain for plant anatomy; counterstain for alum hematoxylin; Kreyberg′s method for keratin and mucus.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Sabnis.
Handbook of Biological Dyes and Stains: Synthesis and Industrial Applications, 544-544 (2010)
R W Sabnis.
Handbook of Acid-Base Indicators (2007)
Farah Maria Drumond Chequer et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3447-3451 (2012-08-01)
Erythrosine (ErB) is a xanthene and an US Food and Drug Administration approved dye used in foods, drugs and cosmetics. Although its utilization is permitted, ErB is described as inhibitor of enzymes and protein-protein interactions and is toxic to pituitary
Laura Carmen Apostol et al.
Biodegradation, 23(5), 725-737 (2012-03-23)
Biodegradation of a xanthene dyes was investigated for the first time using anaerobic granular sludge. On a first screening, biomass was able to decolorize, at different extents, six azo dye solutions: acid orange 7, direct black 19, direct blue 71
Chueh-Pin Chen et al.
Photochemistry and photobiology, 88(3), 570-576 (2012-01-31)
The growing resistance to antibiotics has rendered antimicrobial photodynamic inactivation (PDI) an attractive alternative treatment modality for infectious diseases. Chitosan (CS) was shown to further potentiate the PDI effect of photosensitizers and was therefore used in this study to investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service