Skip to Content
Merck
All Photos(2)

Documents

E5126

Sigma-Aldrich

Etiocholan-3α-ol-17-one

Synonym(s):

3α-Hydroxy-5β-androstan-17-one, 5β-Androsterone, Etiocholanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
2217625
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

Quality Level

form

powder

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

solubility

chloroform: 9.80-10.20 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@@H](O)C[C@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CCC4=O

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13-,14+,15+,16+,18+,19+/m1/s1

InChI key

QGXBDMJGAMFCBF-BNSUEQOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Ganswindt et al.
Physiological and biochemical zoology : PBZ, 85(2), 194-199 (2012-03-16)
Biologically inert material in feces may confound interpretations of noninvasive fecal endocrine data, because it may induce variance related to differences in foraging behavior rather than to differences in endocrine activity. We evaluated two different enzyme immunoassays (EIAs) for the
Istaroxime: is the remedy better than the disease?: Editorial to: "Chronic istaroxine improves cardiac function and heart rate variability in cardiomyopathic hamsters" by Pietro Lo Giudice et al.
Dimitrios Farmakis et al.
Cardiovascular drugs and therapy, 25(2), 115-117 (2011-04-13)
Clinical implications of the 5α-androstanedione pathway for castration-resistant prostate cancer.
Nima Sharifi
Future oncology (London, England), 7(11), 1239-1241 (2011-11-03)
Nina Sneitz et al.
The Journal of steroid biochemistry and molecular biology, 127(3-5), 282-288 (2011-09-09)
Steroids enantiomers are interesting compounds for detailed exploration of drug metabolizing enzymes, such as the UDP-glucuronosyltransferases (UGTs). We have now studied the glucuronidation of the enantiomers of estradiol, androsterone and etiocholanolone by the 19 human UGTs of subfamilies 1A, 2A
Richard Houghton et al.
Bioanalysis, 1(8), 1365-1374 (2010-11-19)
While the regulatory guidelines that describe the validation requirements for small molecules are very comprehensive, they are written primarily for xenobiotic drug molecules. However, the presence of endogenous analyte in control matrix presents an added analytical challenge that must be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service