Skip to Content
Merck
All Photos(1)

Key Documents

D9446

Sigma-Aldrich

L-DOPS

≥98% (HPLC)

Synonym(s):

Droxidopa, L-threo 3,4-Dihydroxyphenylserine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO5
CAS Number:
Molecular Weight:
213.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥3 mg/mL

storage temp.

−20°C

SMILES string

N[C@@H]([C@H](O)c1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO5/c10-7(9(14)15)8(13)4-1-2-5(11)6(12)3-4/h1-3,7-8,11-13H,10H2,(H,14,15)/t7-,8+/m0/s1

InChI key

QXWYKJLNLSIPIN-JGVFFNPUSA-N

General description

L-DOPS, also called L-threo 3,4-Dihydroxyphenylserine or droxidopa is a catecholamine. L-DOPS has many clinical advantages and may be useful for treating norepinephrine deficiency.[1] It is used for treating neurogenic orthostatic hypotension and improves norepinephrine levels.[2] L-DOPS mediates reduction of amyloid plaques and could have a therapeutic potential for treating amyloid pathology.[3]

Application

L-DOPS has been administered for clinical trial studies in mice with amyloid pathology.[3]

Biochem/physiol Actions

L-DOPS is a norepinephrine precursor in vivo.

Features and Benefits

This compound is featured on the Dopamine, Norepinephrine and Epinephrine Synthesis page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Horacio Kaufmann et al.
Neurology, 83(4), 328-335 (2014-06-20)
To determine whether droxidopa, an oral norepinephrine precursor, improves symptomatic neurogenic orthostatic hypotension (nOH). Patients with symptomatic nOH due to Parkinson disease, multiple system atrophy, pure autonomic failure, or nondiabetic autonomic neuropathy underwent open-label droxidopa dose optimization (100-600 mg 3
Jill M Wecht et al.
Archives of physical medicine and rehabilitation, 94(10), 2006-2012 (2013-04-23)
To determine the effect of an escalating dose of droxidopa (100, 200, and 400 mg) compared with placebo on seated blood pressure (BP) in hypotensive individuals with spinal cord injury (SCI). Secondarily, we aimed to determine the effect of droxidopa
Kafui Dzirasa et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(18), 6387-6397 (2010-05-07)
Although normal dopaminergic tone has been shown to be essential for the induction of cortico-striatal and mesolimbic theta oscillatory activity, the influence of norepinephrine on these brain networks remains relatively unknown. To address this question, we simultaneously recorded local field
Horacio Kaufmann
Clinical autonomic research : official journal of the Clinical Autonomic Research Society, 18 Suppl 1, 19-24 (2008-04-23)
Neurogenic orthostatic hypotension results from failure to release norepinephrine, the neurotransmitter of sympathetic postganglionic neurons, appropriately upon standing. In double blind, cross over, placebo controlled trials, administration of droxidopa, a synthetic amino acid that is decarboxylated to norepinephrine by the
Courtney Holmes et al.
Clinical chemistry, 56(5), 832-838 (2010-03-09)
L-threo-3,4-dihydroxyphenylserine (L-DOPS, droxidopa) is a norepinephrine (NE) prodrug under development to treat orthostatic hypotension. 3,4-Dihydroxyphenylacetaldehyde (DOPAL), an endogenous catecholaldehyde produced by enzymatic oxidative deamination of dopamine, is toxic to catecholaminergic neurons. Based on the observation of increasing plasma DOPAL after

Articles

Dopamine-β-hydroxylase is located inside amine storage vesicles of norepinephrine neurons. Dopamine is actively transported from the cytoplasm into the vesicles. As the enzyme is a copper containing protein, its activity can be inhibited by copper chelating agents, such as diethyldithiocarbamate and FLA-63. Inhibition of the enzyme effectively reduces tissue norepinephrine levels.

Dopamine-β-hydroxylase is located inside amine storage vesicles of norepinephrine neurons. Dopamine is actively transported from the cytoplasm into the vesicles. As the enzyme is a copper containing protein, its activity can be inhibited by copper chelating agents, such as diethyldithiocarbamate and FLA-63. Inhibition of the enzyme effectively reduces tissue norepinephrine levels.

Dopamine-β-hydroxylase is located inside amine storage vesicles of norepinephrine neurons. Dopamine is actively transported from the cytoplasm into the vesicles. As the enzyme is a copper containing protein, its activity can be inhibited by copper chelating agents, such as diethyldithiocarbamate and FLA-63. Inhibition of the enzyme effectively reduces tissue norepinephrine levels.

Dopamine-β-hydroxylase is located inside amine storage vesicles of norepinephrine neurons. Dopamine is actively transported from the cytoplasm into the vesicles. As the enzyme is a copper containing protein, its activity can be inhibited by copper chelating agents, such as diethyldithiocarbamate and FLA-63. Inhibition of the enzyme effectively reduces tissue norepinephrine levels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service