Skip to Content
Merck
All Photos(4)

Key Documents

D8764

Sigma-Aldrich

Deoxyribonuclease II from bovine spleen

Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein

Synonym(s):

DNase II, Deoxyribonucleate 3′-oligonucleotido-hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine spleen

type

Type V

form

essentially salt-free, lyophilized powder

specific activity

≥1,000 units/mg protein

mol wt

38 kDa

concentration

≥50% protein

technique(s)

DNA extraction: suitable
tissue culture: suitable

suitability

suitable for molecular biology

application(s)

cell analysis

foreign activity

RNase ≤0.1%

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Deoxyribonuclease II from bovine spleen has been used:
  • as a component of digestion mix to isolate immune cells from colon samples
  • to digest retinal explants for single-cell suspension preparation
  • for isolation of stromal cells from human colon tissue
  • in the dissociation medium during the preparation of embryonic cardiac myocytes from rat heart

Biochem/physiol Actions

Deoxyribonuclease II, also called as acid DNAse, hydrolyzes deoxyribonucleotide linkages in native and denatured DNA yielding products with 3′-phosphates. in vitro, its optimum pH range is 4.5 - 5.0. It also acts upon p-nitrophenyl-phosphodiesters at pH 5.6 - 5.9. The molecular weight is approximately 38,000 Da.

Unit Definition

One Kunitz unit will produce a ΔA260 of 0.001 per min per mL at pH 4.6 at 25 °C; [Mg2+] = 0.83 mM

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Nitta et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 114(2), 119-128 (1994-02-01)
Deoxyribonucleases from eggs and the liver of Xenopus laevis were partially purified by DEAE-cellulose and heparin-Sepharose affinity column chromatographies. The fractions having egg and liver DNase activities were eluted on high performance liquid chromatography through TSK gel G3000SW at the
S Ingebrandt et al.
Biosensors & bioelectronics, 16(7-8), 565-570 (2001-09-07)
An extracellular recording system has been designed for the detection of electrical cell signals using p-channel or n-channel field-effect transistor (FET) arrays with non-metallized gates. Signals from rat heart muscle cell were recorded by these devices and the results described
STUDIES ON ACID DEOXYRIBONUCLEASE. II. ISOLATION AND CHARACTERIZATION OF SPLEEN-ACID DEOXYRIBONUCLEASE.
G BERNARDI et al.
Biochemistry, 3, 1419-1426 (1964-10-01)
Dopamine D2 receptor stimulation differentially affects voltage-activated calcium channels in rat pituitary melanotropic cells.
Keja J A, et al.
The Journal of Physiology, 450(1), 409-435 (1992)
Yanick J Crow
European journal of immunology, 40(9), 2376-2378 (2010-08-14)
DNase II is an endonuclease which plays a fundamental role in the degradation of DNA from both apoptotic cells, and nuclei extruded from red blood cells during erythropoiesis: important tasks, considering that everyday 10(8)-10(9) cells undergo apoptosis, and 10(11) red

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service