D4263
Deoxyribonuclease I from bovine pancreas
Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Synonym(s):
DNase I, Deoxyribonucleate 5′-oligonucleotido-hydrolase
About This Item
Recommended Products
biological source
bovine pancreas
form
powder
specific activity
1700—2300 U/vial
mol wt
~31 kDa
purified by
chromatography
packaging
vial of ≥0.25 mg total protein
technique(s)
DNA extraction: suitable
solubility
0.15 M NaCl: soluble 5.0 mg/mL, clear
suitability
suitable for molecular biology
application(s)
diagnostic assay manufacturing
diagnostic assay manufacturing
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Biochem/physiol Actions
Features and Benefits
Versatile applications: This enzyme can be used to nick DNA, incorporate labeled bases into DNA, and isolate and molecularly characterize cancer stem cells in murine breast tumors. It is also useful in comparative studies of deoxyribonuclease isolated from bovine, ovine, and porcine pancreas.
Stable: The enzyme remains active for up to five hours at 60 °C between pH 5 and 7, and can be stored for up to a week at −20 °C with minimal loss of activity. This stability makes it a cost-effective and convenient option.
Unit Definition
Preparation Note
Analysis Note
inhibitor
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
To standardize a procedure for the enzymatic assay of Deoxyribonuclease I.
To standardize a procedure for the enzymatic assay of Deoxyribonuclease I.
To standardize a procedure for the enzymatic assay of Deoxyribonuclease I.
To standardize a procedure for the enzymatic assay of Deoxyribonuclease I.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service