Skip to Content
Merck
All Photos(3)

Documents

D8035

Sigma-Aldrich

n-Dodecyl β-D-glucopyranoside

≥98% (GC)

Synonym(s):

Dodecyl glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36O6
CAS Number:
Molecular Weight:
348.47
Beilstein:
86236
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Assay

≥98% (GC)

form

powder

mol wt

348.47 g/mol

technique(s)

protein quantification: suitable

CMC

0.13

solubility

methanol: soluble 50 mg/mL, clear to very slightly hazy, colorless

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C18H36O6/c1-2-3-4-5-6-7-8-9-10-11-12-23-18-17(22)16(21)15(20)14(13-19)24-18/h14-22H,2-13H2,1H3/t14-,15-,16+,17-,18-/m1/s1

InChI key

PYIDGJJWBIBVIA-UYTYNIKBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dodecyl β--D-glucopyranoside, a long chain alkyl glycopyranoside and a classical nonionic amphiphile surfactant, is used in detergent and colloid research and micelle development. It may be used as a reference compound in long-chain alkyl glucoside separation and analysis procedures.

Biochem/physiol Actions

Non-ionic saccharide detergent used for the solubilization of membrane-bound proteins, such as G-Protein-Coupled Receptors, in native state

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Daicho et al.
Biochimica et biophysica acta, 1107(1), 61-69 (1992-06-11)
A study of the effects of alkyl glycosides incorporated into synthetic liposomes with respect to their stability, their in vivo distribution in Ehrlich solid tumor-bearing mice and their in vitro interaction with liver cells was undertaken. The synthetic liposomes were
S H El-Sharkawy
Australasian biotechnology, 6(1), 8-12 (1996-01-01)
A number of microorganisms has been screened for growth on sunflower oil as a sole carbon source for production of useful chemicals. Rhizopus stolonifer NRRL 1478 was found to transform the lipid contents of sunflower oil into dodecyl b-D-glucopyranoside and
Caroline A Ericsson et al.
Carbohydrate research, 340(8), 1529-1537 (2005-05-11)
The solid-state phase behaviour of lyophilised n-dodecyl-beta-D-glucoside (beta-C(12)G(1)), n-dodecyl-beta-D-maltoside (beta-C(12)G(2)) and n-dodecyl-beta-D-maltotrioside (beta-C(12)G(3)) has been investigated by differential scanning calorimetry (DSC) and X-ray techniques. For beta-C(12)G(1), lyophilisation results in a formation of a crystalline anhydrate. The lamellar spacing (37 Angstroms)
R R Loo et al.
Protein science : a publication of the Protein Society, 3(11), 1975-1983 (1994-11-01)
Electrospray ionization mass spectrometry (ESI-MS) has proven to be a useful tool for examining noncovalent complexes between proteins and a variety of ligands. It has also been used to distinguish between denatured and refolded forms of proteins. Surfactants are frequently
V Vill et al.
Chemistry and physics of lipids, 104(1), 75-91 (2000-02-05)
A systematic structure variation of a classical amphiphile (dodecyl-beta-D-glucopyranoside) is performed, demonstrating the influence of anomeric linkage, configuration, ring size and flexibility as well as electric charges on the mesophase behaviour. In addition, we have investigated the thermotropic and lyotropic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service