Skip to Content
Merck
All Photos(2)

Key Documents

850510P

Avanti

n-nonyl-β-D-glucoside

n-nonyl-β-D-glucopyranoside, powder

Synonym(s):

nonyl glucoside; NG; NGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O6
CAS Number:
Molecular Weight:
306.40
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (contains <0.2% alpha isomer, TLC)

form

powder

mol wt

306.40 g/mol

packaging

pkg of 1 × 1 g (850510P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand 850510P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCCCCCCCC)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C15H30O6/c1-2-3-4-5-6-7-8-9-20-15-14(19)13(18)12(17)11(10-16)21-15/h11-19H,2-10H2,1H3/t11-,12-,13+,14-,15-/m1/s1

InChI key

QFAPUKLCALRPLH-UXXRCYHCSA-N

General description

n-nonyl-β-D-glucoside is a non-ionic(3) surfactant.[1]
Nonyl glucoside is a dialyzable nonionic detergent for functional solubilization and purification of membrane proteins.

Biochem/physiol Actions

n-nonyl-β-D-glucoside is usually preferred for membrane protein crystallization trials.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (850510P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interaction of n-nonyl-beta-d-glucopyranoside and sodium dodecyl sulfate with DMPC and DMPG monolayers studied by infrared reflection absorption spectroscopy
Meister A, et al.
Physical Chemistry Chemical Physics, 6(24), 5543-5550 (2004)
Tuan A Nguyen et al.
Journal of molecular microbiology and biotechnology, 18(2), 85-91 (2010-02-18)
A cell-free expression system using an Escherichia coli extract was adapted for large-scale expression and purification of mammalian membrane proteins. The system was tested with a set of human membrane proteins of different sizes, numbers of transmembrane domains, oligomeric arrangements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service